Drukuj
Methane in nitrogen, analytical standards Supelco®
Dostawca: Merck
SUPL23443EA 841.5 PLN
SUPL23443 SUPL307300-U
Methane in nitrogen, analytical standards Supelco®
Methane in nitrogen, 1% is a two component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analyzers.