Poland
Order Entry
Supelco® PESTANAL® Organic Reference Standard, 4,4′-DDT
  31041-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, 4,4′-DDT
  31041-100MG
 :  Merck
 :  
 
462 zł
   
  1 * 100 mg

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    50-29-3
  • Description:
    4,4′-DDT
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 + H311 - H351 - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000802
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane; 1,1-Bis(4-chlorophenyl)-2,2,2-trichloroethane
  • Melting point:
    107...110 °C (lit.)
  • Formula:
    (ClC₆H₄)₂CHCCl₃
  • Molecular weight:
    354,49
  • Flash point:
    Not applicable