Supelco® PESTANAL® Organic Reference Standard, Fluazinam
34095-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:79622-59-6
- Description:Fluazinam
- Storage conditions:2...30 °C
- Hazard class description:H317 - H318 - H332 - H361d - H410
- Shipping conditions:STD
- MDL No.:MFCD00214168
- M-Clarity quality level:MQ100
- Synonym(s):3-Chloro-N-(3-chloro-5-trifluoromethyl-2-pyridyl)-α,α,α-trifluoro-2,6-dinitro-p-toluidine; 3-Chloro-N-(3-chloro-2,6-dinitro-4-trifluoromethylphenyl)-5-trifluoromethyl-2-pyridylamine
- Formula:C₁₃H₄Cl₂F₆N₄O₄
- Molecular weight:465,09
- Flash point:Not applicable