Drukuj
Ethylene in helium, analytical standards, Supelco®
Dostawca: Merck
SUPL22572EA 1026 PLN
SUPL22572
Ethylene in helium, analytical standards, Supelco®
Suitable for GC calibration, testing of fixed and personal gas monitors, and testing of in situ industrial stack analysers.
  • A certificate of analysis is included
  • Packaged in lightweight, non-refillable, recyclable containers
  • Portable, convenient, and easy to use
  • Manufactured gravimetrically for maximum accuracy
  • NIST traceable