Drukuj
F.A.M.E. Mix GLC-80, certified reference material, TraceCERT®, Supelco®
Dostawca: Merck
TraceCERT®
1898-1AMPEA 272 PLN
1898-1AMP
F.A.M.E. Mix GLC-80, certified reference material, TraceCERT®, Supelco®
Standardy Wzorce żywności
Methyl heptadecanoate 20 wt. %; Methyl myristate 20 wt. %; Methyl palmitate; Methyl pentadecanoate 20 wt. %; Methyl tridecanoate 20 wt. %
Certyfikaty: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.