Drukuj
Nitrogen generator, Genius AB-3G - SCIEX
Dostawca: PEAK SCIENTIFIC
PEAK10-8010EA 0 PLN
PEAK10-8010
Nitrogen generator, Genius AB-3G - SCIEX
Generatory gazu Generatory azotu
The AB-3G has been designed to operate to the exact requirements of several Sciex LC-MS instruments. A three gas system, the AB-3G provides nitrogen curtain gas, dry air source gas and dry air exhaust gas and benefits from robust integrated compressors, making it a complete standalone solution.
  • Built to ensure uninterrupted supply of gas for clinical applications
  • Advanced alarms in the unlikely event of product issues
  • Generator has the capacity to self-diagnose and inform when maintenance is required
  • Highly economical source of nitrogen gas with low lifetime running costs
  • Compressors are housed in an insulated compartment to reduce noise and vibration
  • 1 year comprehensive on-site warranty
These are self-contained systems featuring integrated compressors, delivering laboratory-grade nitrogen at a range of pressures and flow rates to satisfy the requirements of a wide selection of instruments.
Current: 9,6 A
Outlet pressure:
6,2 bar (Nitrogen, dry air)
7,5 bar/4,1 bar (dry air)
Certyfikaty: Conforms with: Low Voltage Directive 2014/35/EU; EN 61010-1: 2010 Electrical Equipment for measurement, control and laboratory use.
Electromagnetic Compatibility Directive 2014/130/EU; EN 61326-1: 2013 Electrical Equipment for measurement, control and laboratory use.
FCC47 CFR Part 15 Class B.
Unintentional radiators; Conducted and Radiated emissions limits.