Drukuj
Omegawax® column test mix, analytical standard, Supelco®
Dostawca: Merck
48476.EA 485 PLN
48476.
Omegawax® column test mix, analytical standard, Supelco®
Standardy Wzorce do chromatografii
Omegawax® column test mix has been used as an standard in studying the effect of supplementation with procyanidins obtained from Vitis vinifera seeds on certain selected markers of oxidative stress, via the analysis of fatty acid composition of phospholipids of red blood cell membrane using gas chromatography method of analysis (GC).