Drukuj
Avantor® Hichrom Pellicular ODS, kolumny do HPLC
Dostawca: VWR
PEODS-20DKEA 0 PLN
PEODS-20DK PEODS-125AM PEODS-50G PEODS-75GM PEODS-10C5 PEODS-20CA3 HICHPEODS-150AM
Avantor® Hichrom Pellicular ODS, kolumny do HPLC
Kolumny chromatograficzne
Pellicular ODS columns are designed to provide high resolution and efficiency for separations of protein variants.
Octadecylsilane chemically bonded to silica gel of a controlled surface porosity that has been bonded to a solid spherical core, 30 to 50 μm in diameter.