LiChrospher® RP-Select B, HPLC Columns
Supplier: Avantor
Certificates
About this item
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- Base deactivated LiChrospher® RP-select B
- High efficiency packed columns
Specifications
- Catalog No:
- LISPRPB-5-150A
- LISPRPB-5-250A
- LISPRPB-5-10C5
- LISPRPB-5-125AF
- LISPRPB-5-125A
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 11.5%
- 11.5%
- 11.5%
- 11,5%
- 11.5%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 360 m²/g
- 360 m²/g
- 360 m²/g
- 360 m²/g
- 360 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
LiChrospher® Analytical columns
Product Information
- Catalog #PhaseColumn FormatColumn hardwarePore sizeParticle SizeØ int.LengthAvailabilityPrice
- LISPRPB-5-150ARP-Select BStainless steel60 Å5 µm4,60 mm150 mmLog in for availability1 * 1 SZT5 320 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load11.5%EndcappedYesSurface area360 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - LISPRPB-5-250ARP-Select BStainless steel60 Å5 µm4,60 mm250 mmLog in for availability1 * 1 SZT5 550 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load11.5%EndcappedYesSurface area360 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - LISPRPB-5-125AFRP-Select BStainless steel60 Å5 µm4,00 mm125 mmLog in for availability1 * 1 SZT4 750 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load11,5%EndcappedYesSurface area360 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - LISPRPB-5-125ARP-Select BStainless steel60 Å5 µm4,60 mm125 mmLog in for availability1 * 1 SZT5 220 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load11.5%EndcappedYesSurface area360 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
LiChrospher® Guard cartridges
Product Information
- Catalog #PhaseColumn FormatColumn hardwarePore sizeParticle SizeØ int.LengthAvailabilityPrice
- LISPRPB-5-10C5RP-Select BGuard cartridgeStainless steel60 Å5 µm3,20 mm10 mmLog in for availability1 * 5 SZT1 400 złSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load11.5%EndcappedYesSurface area360 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact