Order Entry
Scrub suit, SMS
Scrub suit, SMS
undefined
Scrub suit, SMS
  •  
    Colour
    Clothing Size
    Pk
  • Scrub suit, dark blue, 40 g/m²
    Scrub suit, dark blue, 40 g/m²
    HOIT119724
    Dark blue
    XL
    50
    1 * 50 SZT
    1 * 50 SZT - 1 130 zł
    1 130 zł
    1 * 50 SZT
       1 130 zł
     : HOIT119724
    Specifications
    Environmentally Preferable
    Gender
    Garment application
  • Scrub suit, white, 45 g/m²
    Scrub suit, white, 45 g/m²
    HOIT121934
    White
    L
    50
    1 * 50 SZT
    1 * 50 SZT - 1 270 zł
    1 270 zł
    1 * 50 SZT
       1 270 zł
     : HOIT121934
    Specifications
    Environmentally Preferable
    Gender
    Garment application