Drukuj
Wyszukano: Chusteczki do ogólnych zastosowań
51  wyniki/ów wyszukiwania
SearchResultCount:"51"
Sort Results
Widok listy Widok uproszczony (nowość)
Oceń wyniki wyszukiwania
Producent: VWR Collection
Opis: Chusteczki wykonane w technice spunlace z 55% pulpy drzewnej i 45% poliestru są wyjątkowo mocne, nawet gdy są mokre.
Numer katalogowy: (115-2418)
Producent: VWR Collection
Opis: These melt-blown wipes, made from 100% PP, have a very high absorption capacity.
j.m.: 1 * 1 Roll
Producent: VWR Collection
Opis: These spunlace apertured wipes, made from 51% wood pulp and 49% polyester, are slightly abrasive on one side and soft on the other.
Wyprzedaż
Numer katalogowy: (115-2417)
Producent: VWR Collection
Opis: These very soft wipes, made from a spunlace fabric, consisting of 60% viscose and 40% polyester, are ideal for sensitive surfaces and light duty clean...
j.m.: 1 * 500 SZT
Producent: VWR Collection
Opis: Te wybielone chusteczki wykonane w technice spunlace z 55% pulpy drzewnej i 45% poliestru są wyjątkowo mocne, nawet gdy są mokre.
Producent: PGI NONWOVEN
Opis: These semi-reusable wipes are perfectly suited for precision cleaning and critical wiping tasks across a range of industries. Thanks to their composit...
Numer katalogowy: (115-2051)
Producent: KIMBERLY CLARK
Opis: These single ply wipes are made from AIRFLEX* material, a specialised fabric with a perfect balance of softness and strength, that is also thick and a...
j.m.: 1 * 1 Roll
Producent: KIMBERLY CLARK
Opis: These single-use wipes, made from two ply AIRFLEX* material, are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass,...
Producent: KIMBERLY CLARK
Opis: WypAll® L10 professional wiping paper products are designed for the specific environments where they will be used, helping enable you to deliver on yo...
Producent: KIMBERLY CLARK
Opis: These single use wipes made from single ply AIRFLEX* material are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass...
Producent: PGI NONWOVEN
Opis: Microfibre Light cloths provide the same superior cleaning performance as traditional woven microfibre cloths without the risk of cross contamination....
Producent: KIMBERLY CLARK
Opis: These single use wipes made from three ply AIRFLEX* material are ideal for superior cleaning and heavy duty wiping tasks such as soaking up industrial...
Numer katalogowy: (KIMB72773)
Producent: KIMBERLY CLARK
Opis: WypAll® wiping paper products are designed for the specific environments where they will be used, to deliver on your wiping task, every time.
j.m.: 1 * 6 Roll
Producent: KIMBERLY CLARK
Opis: Due to its HYDROKNIT* material, WYPALL* X50 cloth provides fast absorption, tear resistance and high performance.
Producent: KIMBERLY CLARK
Opis: WYPALL* L40 wipes easily absorb large spills and leaks and are therefore ideal for routine industrial cleaning and maintenance. They are also soft eno...
Producent: KIMBERLY CLARK
Opis: These soft two or three layer sheets are designed for easy underlaying for outpatient examinations or treatments. Ideal for use in hospitals, clinics,...
Cena wymaga potwierdzenia u dostawcy
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Zapasy tego artykułu są ograniczone, ale możliwe, że jest on dostępny w pobliskim magazynie. Upewnij się, że jesteś zalogowany na stronie, aby móc sprawdzić dostępność zapasów. Jeśli call ciągle się wyświetla i potrzebujesz pomocy, zadzwoń na 58 323 82 00.
Ten produkt jest ograniczony w dostępie i można go zamówić tylko przy użyciu zaaprobowanego konta. Jeśli potrzebujesz pomocy, napisz do Działu Prawnego VWR na tomasz.chrobak@pl.vwr.com
Do zakupu tego produktu może być potrzebna dodatkowa dokumentacja. Przedstawiciel VWR skontaktuje się z Tobą w razie potrzeby.
Ten produkt jest zablokowany na stronie internetowej. W celu zamówienia, skontaktuj się z Działem Obsługi Klienta.
Produkt oryginalny został wycofany. Dostępny jest wskazny zamiennik .
Wybrany produkt został wycofany - sprzedaż do wyczerpania zapasów. Dostępne zamienniki można znaleźć poprzez wpisanie powyższego numeru katalogowego VWR w okno wyszukiwarki. Jeśli potrzebujesz dalszej pomocy, prosimy o kontakt z Działem Doradztwa Produktowego pod numerem telefonu 58 3238 220.
1 - 16 of 51
no targeter for Bottom