Order Entry
VWR® ARISTAR®, Boron Standard Solution, 1000 mg/L B in Ammonium Hydroxide Solution (max. 1%) (from B(OH₃)), Standard for ICP
Supplier:  VWR Chemicals
Certificates
undefined
VWR® ARISTAR®, Boron Standard Solution, 1000 mg/L B in Ammonium Hydroxide Solution (max. 1%) (from B(OH₃)), Standard for ICP
Product Information
  • Catalog #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • Boron
    B
    1000 µg/ml
    <1% NH₄OH
    Plastic bottle
    500 mL
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 450 zł
    1 450 zł
    1 * 500 mL
       1 450 zł
    Local Article Number: VWRC455124L
  • Boron
    B
    1000 µg/ml
    <1% NH₄OH
    Plastic bottle
    100 mL
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 731 zł
    731 zł
    1 * 100 mL
       731 zł
    Local Article Number: VWRC455122J