Drukuj
Etykiety, przywieszki, etykiety bezpieczeństwa
Dostawca: Brady
BRDY225256EA 104 PLN
BRDY225256 BRDY241716 BRDY241718
Etykiety, przywieszki, etykiety bezpieczeństwa
Etykiety
Provide on-the-spot identification of safety measures.
  • Easy application: can be attached in just second
  • Tags have ties attached and can be applied to just about any part of any machine
  • The tag is printed on both sides, making sure the vital information is always visible
  • Material: Heavy duty polyester 0,25 mm thickness
Size of attachment tie: 15×140 mm