Avantor® Hichrom HI-OVW, GC Columns
Supplier: Avantor
Certificates
About this item
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI94-32-015-15
- HI94-32-010-15
- Phase name:
- HI-OVW
- HI-OVW
- Phase composition:
- Proprietary Phase developed for Waxes Analysis in Edible Oils
- Proprietary Phase developed for Waxes Analysis in Edible Oils
- Crossbond:
- Yes
- Yes
Specifications
Product Information
- Catalog #Column FormatFilm thicknessØ int.LengthPkAvailabilityPrice
- GC columnGC columnHI94-32-015-15Capillary0,10 - 0,15 µm0,32 mm15 m1Log in for availability1 * 1 SZT4 650 złLocal Article Number: HIGCHI94-32-015-15Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-OVWPhase compositionProprietary Phase developed for Waxes Analysis in Edible OilsCrossbondYes - GC columnLocal Article Number: HIGCHI94-32-010-15Specifications:
More
Accessories
Specifications
Environmentally PreferablePhase nameHI-OVWPhase compositionProprietary Phase developed for Waxes Analysis in Edible OilsCrossbondYes