Order Entry
Lead ICP standards, (CRMs), Fluka™
Lead ICP standards, (CRMs), Fluka™
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • HONCE1170-100ML
    E1170-100ML
    Lead
    Pb
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 691 zł
    691 zł
    1 * 100 mL
       691 zł
  • HONCE1043-100ML
    E1043-100ML
    Lead
    Pb
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 484 zł
    484 zł
    1 * 100 mL
       484 zł