Order Entry
Praseodymium ICP & ICP-MS Standards, (CRMs), Fluka™
Praseodymium ICP & ICP-MS Standards, (CRMs), Fluka™
Certificates
Praseodymium ICP & ICP-MS Standards, (CRMs), Fluka™
Product Information
  • Catalog #
    Supplier #
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
    Availability
    Price
  • HONCE1341-100ML
    E1341-100ML
    Praseodymium
    Pr
    100 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 553 zł
    553 zł
    1 * 100 mL
       553 zł
    Supplier Article Number: E1341-100ML
    Local Article Number: HONCE1341-100ML
  • HONCE1409-100ML
    E1409-100ML
    Praseodymium
    Pr
    10 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 471 zł
    471 zł
    1 * 100 mL
       471 zł
    Supplier Article Number: E1409-100ML
    Local Article Number: HONCE1409-100ML