Order Entry
Rubidium ICP and ICP-MS Standards, (CRMs), Fluka™
Rubidium ICP and ICP-MS Standards, (CRMs), Fluka™
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • HONCE1343-100ML
    E1343-100ML
    Rubidium
    Rb
    100 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 484 zł
    484 zł
    1 * 100 mL
       484 zł
  • HONCE1411-100ML
    E1411-100ML
    Rubidium
    Rb
    10 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 471 zł
    471 zł
    1 * 100 mL
       471 zł