Drukuj
Termostaty grzewcze z transparentną łaźnią, ECO
Dostawca: Lauda
ECO
461-1236EA 12140 PLN
461-1236 LAUDL001128 LAUDL001131 LAUDL001132 LAUDL001127 LAUDL001130 LAUDL001125 461-1289 461-1239 461-1240 LAUDL001098 LAUDL001157 LAUDL001129 LAUDL001161 461-1237 LAUDL001158 461-1238 LAUDL001160 461-1288 LAUDL001155 LAUDL001154 LAUDL001159 LAUDL001156 LAUDL001126
Termostaty grzewcze z transparentną łaźnią, ECO
Termostaty cyrkulacyjne Systemy cyrkulacyjne, otwarte Łaźnie ze zdejmowanym cyrkulatorem
ECO units with transparent plastic baths provide visibility in all cases where test samples need to be observed during thermostating. The thermostats with polycarbonate baths can be used in the temperature range of up to 100 °C. They have a filling volume of 5 up to 20 litres. The 15 litre (or larger) Silver and Gold baths accommodate up to three capillary viscometers. The Silver and Gold ET 6, ET 12 and ET 20 models are equipped with a cooling coil as standard. Bath/circulation thermostats have menu navigation in plain text for user friendly operation. Various interface modules are available as optional accessories.
  • Variopump with six adjustable stages
  • Flow rate switch for flow rate control between internal and external circulation
  • Simultaneous display of set and actual temperature
  • Over-temperature setting via menu
  • USB interface as standard
Silver controller.
Heating power 2 kW, working temperature range up to 100 °C
LCD display, resolution of 0,01 °C
Programmer with one program and 20 segments
Gold controller
Heating power 2,6 kW, working temperature range up to 100 °C
Colour TFT display, resolution of 0,01 °C
Programmer with five programs and 150 segments
Graphical display of temperature profiles
Ramp function and clock timer
Maximum pump capacity:
All models: 22 L/min, 0,55 bar
Informacje dodatkowe: Supplied with cooling coil (except ET 15 S and ET 15 G models). The ET 15 S model includes a pump circulation set with 13 mm diameter plastic nipples as standard, ET 15 G includes a pump circulation set with M16×1 threads. A wide range of optional modules and accessories such as covers for  15 litre models are available, please enquire for details.