VWR® Semi-Micro Balances, SM Classic: LCD Display
Supplier: VWR Collection
Certificates
LCD display models without factory calibration certificates
Product Information
- Catalog #ModelWeighing cap.ReadabilityPan/Platform SizeLinearityPkAvailabilityPrice
- 611-3534SM 1265Di62/120 g0,01/0,1 mgØ 80 mm±0,08/±0,2 mg11 * 1 SZT9 700 złLocal Article Number: VWRI611-3534Specifications:
Accessories
Specifications
Environmentally Preferable - 611-3537SM 2285Di82/220 g0,01/0,1 mgØ 80 mm±0,1/±0,2 mg11 * 1 SZT11 510 złLocal Article Number: VWRI611-3537Specifications:
Accessories
Specifications
Environmentally Preferable - 611-3547SM 425i42 g0,01 mgØ 80 mm±0,05 mg11 * 1 SZT8 130 złLocal Article Number: VWRI611-3547Specifications:
Accessories
Specifications
Environmentally Preferable
LCD balance with calibration certificate issued by an ISO/IEC 1702 accredited laboratory
Product Information
- Catalog #ModelWeighing cap.ReadabilityPan/Platform SizeLinearityPkAvailabilityPrice
- 611-5614SM 2285Di-C82/220 g0,01/0,1 mgØ 80 mm±0,1/±0,2 mg11 * 1 SZT12 390 złLocal Article Number: VWRI611-5614Specifications:
Accessories
Specifications
Environmentally Preferable
LCD display models with factory calibration certificates
Product Information
- Catalog #ModelWeighing cap.ReadabilityPan/Platform SizeLinearityPkAvailabilityPrice
- 611-3539SM 1265Di-C62/120 g0,01/0,1 mgØ 80 mm±0,08/±0,2 mg11 * 1 SZT10 190 złLocal Article Number: VWRI611-3539Specifications:
Accessories
Specifications
Environmentally Preferable - 611-3549SM 425i-C42 g0,01 mgØ 80 mm±0,05 mg11 * 1 SZT8 720 złLocal Article Number: VWRI611-3549Specifications:
Accessories
Specifications
Environmentally Preferable - 611-3542SM 2285Di-C82/220 g0,01/0,1 mgØ 80 mm±0,1/±0,2 mg11 * 1 SZT12 000 złLocal Article Number: VWRI611-3542Specifications:
Accessories
Specifications
Environmentally Preferable