Order Entry
Water purification systems, Milli-Q® IQ 7003/05/10/15
 :  Merck
undefined
Water purification systems, Milli-Q® IQ 7003/05/10/15
 :  Merck
Specifications
  • Catalog No:
  • 171-1379
  • 171-1378
  • 171-1380
  • 171-1381
  • Model:
  • Milli-Q® IQ 7005
  • Milli-Q® IQ 7003
  • Milli-Q® IQ 7010
  • Milli-Q® IQ 7015
  • Feed water:
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Production flow rate:
  • 5 L/h
  • 3 L/h
  • 10 L/h
  • 15 L/h
  • TOC content:
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
  • Resistivity at +25 °C:
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • 18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
  • Bacteria:
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • <0,01 cfu/ml (Type 1, from Q-POD® dispenser);
    <0,01 cfu/ml (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® or Biopak® filter when installed and used in a laminar flow hood; values are typical & may vary with the feed water)
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
  • Particles:
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • Endotoxin:
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • RNases:
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
  • DNases:
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
  • W×D×H:
  • 315×380×498 mm
  • 315×380×498 mm
  • 375×380×498 mm
  • 375×380×498 mm
  • Weight:
  • 26 kg
  • 26 kg
  • 29,7 kg
  • 30 kg
Specifications
  •  
    Pk
  • Milli-Q® IQ 7003 purification system, 3 L/h flow rate
    Milli-Q® IQ 7003 purification system, 3 L/h flow rate
    171-1378
    ZIQ7003T0C
    1
    1 * 1 SZT
    1 * 1 SZT - 75 600 zł
    75 600 zł
    1 * 1 SZT
       75 600 zł
    Accessories
    Specifications
    Model
    Milli-Q® IQ 7003
    Environmentally Preferable
    Feed water
    Potable tap water (consult manual for requirements)
    Production flow rate
    3 L/h
    TOC content
    ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
    Resistivity at +25 °C
    18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
    Bacteria
    <0,01 cfu/ml (Type 1, from Q-POD® dispenser);
    <0,01 cfu/ml (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® or Biopak® filter when installed and used in a laminar flow hood; values are typical & may vary with the feed water)
    Particles
    No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    Endotoxin
    <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    RNases
    <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    DNases
    <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
    W×D×H
    315×380×498 mm
    Weight
    26 kg
  • Milli-Q® IQ 7005 purification system, 5 L/h flow rate
    Milli-Q® IQ 7005 purification system, 5 L/h flow rate
    171-1379
    ZIQ7005T0C
    1
    1 * 1 SZT
    1 * 1 SZT - 87 350 zł
    87 350 zł
    1 * 1 SZT
       87 350 zł
    Accessories
    Specifications
    Model
    Milli-Q® IQ 7005
    Environmentally Preferable
    Feed water
    Potable tap water (consult manual for requirements)
    Production flow rate
    5 L/h
    TOC content
    ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
    Resistivity at +25 °C
    18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
    Bacteria
    <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
    Particles
    No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    Endotoxin
    <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    RNases
    <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    DNases
    <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
    W×D×H
    315×380×498 mm
    Weight
    26 kg
  • Milli-Q® IQ 7010 purification system, 10 L/h flow rate
    Milli-Q® IQ 7010 purification system, 10 L/h flow rate
    171-1380
    ZIQ7010T0C
    1
    1 * 1 SZT
    1 * 1 SZT - 99 580 zł
    99 580 zł
    1 * 1 SZT
       99 580 zł
    Accessories
    Specifications
    Model
    Milli-Q® IQ 7010
    Environmentally Preferable
    Feed water
    Potable tap water (consult manual for requirements)
    Production flow rate
    10 L/h
    TOC content
    ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
    Resistivity at +25 °C
    18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
    Bacteria
    <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
    Particles
    No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    Endotoxin
    <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    RNases
    <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    DNases
    <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
    W×D×H
    375×380×498 mm
    Weight
    29,7 kg
  • Milli-Q® IQ 7015 purification system, 15 L/h flow rate
    Milli-Q® IQ 7015 purification system, 15 L/h flow rate
    171-1381
    ZIQ7015T0C
    1
    1 * 1 SZT
    1 * 1 SZT - 113 520 zł
    113 520 zł
    1 * 1 SZT
       113 520 zł
    Accessories
    Specifications
    Model
    Milli-Q® IQ 7015
    Environmentally Preferable
    Feed water
    Potable tap water (consult manual for requirements)
    Production flow rate
    15 L/h
    TOC content
    ≤2 ppb (µg/L); typically ≤5 ppb (Type 1, from Q-POD® dispenser; in the appropriate operating conditions, otherwise typically ≤ 5 ppb.);
    ≤30 ppb (Type 2);
    (values are typical and may vary depending on the feed water)
    Resistivity at +25 °C
    18,2 MΩ·cm (Type 1);
    >5 MΩ·cm; typically 10 - 15 MΩ·cm (Type 2);
    (resistivity can also be displayed non-temperature-compensated as required by USP)
    Bacteria
    <0,01 KBE/ml (Typ1 aus Q-POD®-Spender)
    <0,01 KBE/ml (Typ2 aus E-POD®-Spender+Endfilter)
    Mit Millipak®- oder Biopak®-Filter bei Installation + Verwendung in einer Laminar-Flow-Haube
    Werte sind typisch und können je nach Speisewasser variieren
    Particles
    No particles with size >0,22 µm (Type 1, from Q-POD® dispenser);
    No particles with size >0.22 µm (Type 2, from E-POD® dispenser with final filter);
    (With Millipak® filter, with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    Endotoxin
    <0,001 EU/mL(Type 1, from Q-POD® dispenser);
    <0,001 EU/ml (Type 2, from an E-POD® dispenser with final filter);
    (With Biopak® polisher when installed and used in a laminar flow hood; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    RNases
    <1 pg/ml (Type 1, from Q-POD® dispenser);
    <1 pg/ml (Type 2, from an E-POD® dispenser with final filter);
    (with Biopak® polisher; values are typical and may vary depending on the nature and concentration of contaminants in the feed water)
    DNases
    <5 pg/mL (Type 1, from Q-POD® dispenser)
    <5 pg/ml (Type 2, from an E-POD® dispenser with final filter)
    (With Biopak® polisher; values are typical & may vary with the feed water)
    W×D×H
    375×380×498 mm
    Weight
    30 kg
Consumables
  •  
    Seq
    Description
    Pk
  • 17
    Milli-Q IQ 7010-15 Purification kit, contains IPAK Gard® cartridge and tank vent filter for optimal protection of reverse osmosis (RO) membrane
    1
    1 * 1 SZT
    1 * 1 SZT - 0 zł
    0 zł
  •  
    Seq
    Description
    Pk
  • 13
    IPAK Quanta® polishing cartridge
    1
    1 * 1 SZT
    1 * 1 SZT - 2 970 zł
    2 970 zł
  •  
    Seq
    Description
    Pk
  • 03
    ech2o oxidation lamp (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 3 520 zł
    3 520 zł
  •  
    Seq
    Description
    Pk
  • 05
    ech2o® bactericidal lamp
    1
    1 * 1 SZT
    1 * 1 SZT - 1 770 zł
    1 770 zł
  •  
    Seq
    Description
    Pk
  • 07
    FI load pack
    1
    1 * 1 SZT
    1 * 1 SZT - 3 760 zł
    3 760 zł
  •  
    Seq
    Description
    Pk
  • 10
    IPAK Gard® pretreatment pack
    1
    1 * 1 SZT
    1 * 1 SZT - 0 zł
    0 zł
  •  
    Seq
    Description
    Pk
  • 11
    IPAK Gard® pretreatment pack for hard water
    1
    1 * 1 SZT
    1 * 1 SZT - 5 240 zł
    5 240 zł
  •  
    Seq
    Description
    Pk
  • 01
    Biopak® polisher (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 1 810 zł
    1 810 zł
  •  
    Seq
    Description
    Pk
  • 22
    Vent filter
    1
    1 * 1 SZT
    1 * 1 SZT - 2 190 zł
    2 190 zł
  •  
    Seq
    Description
    Pk
  • 24
    VOC-Pak® polisher (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 2 350 zł
    2 350 zł
  •  
    Seq
    Description
    Pk
  • 06
    EDS-Pak® polisher (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 2 400 zł
    2 400 zł
  •  
    Seq
    Description
    Pk
  • 23
    Vent filter HF (for high-flow applications)
    1
    1 * 1 SZT
    1 * 1 SZT - 1 510 zł
    1 510 zł
  •  
    Seq
    Description
    Pk
  • 04
    ech2o® ASM lamp
    1
    1 * 1 SZT
    1 * 1 SZT - 1 850 zł
    1 850 zł
  •  
    Seq
    Description
    Pk
  • 14
    LC-Pak® polisher (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 3 960 zł
    3 960 zł
  •  
    Seq
    Description
    Pk
  • 02
    ech2o A10 TOC lamp (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 4 070 zł
    4 070 zł
  •  
    Seq
    Description
    Pk
  • 20
    Millipak® 0,22 μm filter (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 1 220 zł
    1 220 zł
  •  
    Seq
    Description
    Pk
  • 09
    IPAK Gard® H 10-15 pretreatment pack
    1
    1 * 1 SZT
    1 * 1 SZT - 5 820 zł
    5 820 zł
  •  
    Seq
    Description
    Pk
  • 16
    Milli-Q IQ 7003-5 Purification kit, contains IPAK Gard® cartridge and tank vent filter for optimal protection of reverse osmosis (RO) membrane
    1
    1 * 1 SZT
    1 * 1 SZT - 0 zł
    0 zł
  •  
    Seq
    Description
    Pk
  • 21
    Millipak® Gold 0,22 μm sterile filter (also compatible with Milli-Q® IQ 7000)
    1
    1 * 1 SZT
    1 * 1 SZT - 1 310 zł
    1 310 zł
  •  
    Seq
    Description
    Pk
  • 08
    IPAK Gard® 10-15 pretreatment pack
    1
    1 * 1 SZT
    1 * 1 SZT - 0 zł
    0 zł