Drukuj

Antiseptic hand sanitising foam, PURELL®

Dostawca: Gojo
PURELL®
GOJO1304-03-EEU00EA 121 PLN
GOJO1304-03-EEU00
Antiseptic hand sanitising foam, PURELL®
Pielęgnacja skóry Środki do dezynfekcji dłoni Płyny i pianki do dezynfekcji skóry
Refill for PURELL® LTX-7™ dispenser.

  • Thick, luxurious foam
  • Efficacious dose of 1,1 ml
  • Made with 100% naturally renewable ethanol
  • Biodegradable formula

Contains 72% v/v absolute ethanol and skin conditioners, kills more than 99,99% of most common germs that may cause illness.

SANITARY SEALED™ refill helps prevent contamination.

Certyfikaty: Certified by Ecospecifier Global GreenTag™.

Uwaga: Use biocides safely. Always read the label and product information before use.
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR