Drukuj

Nafion® R-1100 membrane, sulfonyl fluoride form, Thickness: 0.05 mm

Dostawca: Thermo Scientific

46324.RYEA 0 PLN
46324.RY 46324.FF
Nafion® R-1100 membrane, sulfonyl fluoride form, Thickness: 0.05 mm
Nafion® sulphonylfluoride form
Formuła: C₉F₁₈O₄S
MW: 546,126 g/mol
Temperatura przechowywania: Temperatura otoczenia
Numer MDL: MFCD05665455
Numer CAS: 26654-97-7

Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR