Europium(III) chloride hydrate 99,99%
Supplier: Apollo Scientific
Certificates
About this item
:13759-92-7
Formula:Cl₃Eu·xH₂O
MDL Number:MFCD00217047
:258.32 g/mol (anhy)
Melting Pt:632 °C (dec.)
Density:4.89 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: IN1768-BULKLocal Article Number: APOSIN1768-BULK
- Supplier Article Number: IN1768-10GLocal Article Number: APOSIN1768-10G