Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
Methyl 3-acetylbenzoate
Methyl 3-acetylbenzoate
Methyl 3-acetylbenzoate
Irritant
Methyl 3-acetylbenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 3-acetylbenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
21860-07-1
:
Formula:
C₁₀H₁₀O₃
MDL Number:
MFCD09475799
:
178.185 g/mol
Melting Pt:
43…46 °C
:
GHS07
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR40606-1G
OR40606-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 110 zł
110 zł
-
+
Add to cart
1 * 1 g
110 zł
-
+
Add to cart
APOSOR40606-5G
OR40606-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 414 zł
414 zł
-
+
Add to cart
1 * 5 g
414 zł
-
+
Add to cart
APOSOR40606-10G
OR40606-10G
Bottle
10 g
Log in
for availability
Log in
for availability
1 * 10 g
1 * 10 g - 830 zł
830 zł
-
+
Add to cart
1 * 10 g
830 zł
-
+
Add to cart