4,8-Dichloro-7-fluoroquinoline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1065093-08-4
:
Formula:C₉H₄Cl₂FN
MDL Number:MFCD28010388
:216.039 g/mol
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC28246-250MGLocal Article Number: APOSPC28246-250MG
- Supplier Article Number: PC28246-1GLocal Article Number: APOSPC28246-1G