Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
2-Amino-5-hydroxybenzoic acid
2-Amino-5-hydroxybenzoic acid 95%
2-Amino-5-hydroxybenzoic acid 95%
Irritant
2-Amino-5-hydroxybenzoic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-5-hydroxybenzoic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
394-31-0
:
Formula:
C₇H₇NO₃
MDL Number:
MFCD00007870
:
153.14 g/mol
Melting Pt:
247 °C (dec.)
:
GHS07
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSOR10133-25G
OR10133-25G
Bottle
25 g
Log in
for availability
Log in
for availability
1 * 25 g
1 * 25 g - 116 zł
116 zł
-
+
Add to cart
1 * 25 g
116 zł
-
+
Add to cart
APOSOR10133-100G
OR10133-100G
Bottle
100 g
Log in
for availability
Log in
for availability
1 * 100 g
1 * 100 g - 327 zł
327 zł
-
+
Add to cart
1 * 100 g
327 zł
-
+
Add to cart