2-Bromo-4-chlorothieno[3,2-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:28948-61-0
:
Formula:C₇H₃BrClNS
Melting Pt:119 °C
:GHS07
Storage Temperature:Refrigerator
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: OR59871-250MGLocal Article Number: APOSOR59871-250MG
- Supplier Article Number: OR59871-1GLocal Article Number: APOSOR59871-1G