Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
4-Bromo-3-fluorobenzaldehyde 97%
:
Apollo Scientific
4-Bromo-3-fluorobenzaldehyde 97%
:
Apollo Scientific
:
133059-43-5
:
:
C₇H₄BrFO
:
MFCD03095000
:
203.01 g/mol
:
55…59 °C
:
>110 °C
:
GHS07
:
Refrigerator
Pack type
Pk
APOSPC10596-5G
PC10596-5G
Bottle
5 g
1 * 5 g
1 * 5 g - 110 zł
110 zł
-
+
1 * 5 g
110 zł
-
+
: PC10596-5G
: APOSPC10596-5G
APOSPC10596-25G
PC10596-25G
Bottle
25 g
1 * 25 g
1 * 25 g - 255 zł
255 zł
-
+
1 * 25 g
255 zł
-
+
: PC10596-25G
: APOSPC10596-25G