2-Chloro-4-fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84194-36-5
:
Formula:C₇H₄ClFO
MDL Number:MFCD00042527
:158.56 g/mol
Boiling Pt:118…120 °C/50mmHg
Melting Pt:59…64 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC1828D-25GLocal Article Number: APOSPC1828D-25G
- Supplier Article Number: PC1828D-100GLocal Article Number: APOSPC1828D-100G
- Supplier Article Number: PC1828D-500GLocal Article Number: APOSPC1828D-500G