Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
11H-Perfluoroundecanoic acid
11H-Perfluoroundecanoic acid 97%
11H-Perfluoroundecanoic acid 97%
11H-Perfluoroundecanoic acid 97%
:
Apollo Scientific
11H-Perfluoroundecanoic acid 97%
:
Apollo Scientific
:
1765-48-6
:
:
C₁₁H₂F₂₀O₂
:
MFCD00042431
:
546.11 g/mol
:
100…101 °C
:
3261
:
8,II
:
GHS05
:
GHS07
:
Ambient
Pack type
Pk
APOSPC3148-1G
PC3148-1G
Bottle
1 g
1 * 1 g
1 * 1 g - 363 zł
363 zł
-
+
1 * 1 g
363 zł
-
+
APOSPC3148-5G
PC3148-5G
Bottle
5 g
1 * 5 g
1 * 5 g - 656 zł
656 zł
-
+
1 * 5 g
656 zł
-
+
APOSPC3148-25G
PC3148-25G
Bottle
25 g
1 * 25 g
-
+
1 * 25 g
-
+