Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Poland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0 zł
Order Entry
Home
2,3-Difluoro-4-methoxybenzamide
2,3-Difluoro-4-methoxybenzamide 98%
2,3-Difluoro-4-methoxybenzamide 98%
2,3-Difluoro-4-methoxybenzamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Difluoro-4-methoxybenzamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
886500-67-0
:
Formula:
C₈H₇F₂NO₂
MDL Number:
MFCD04115983
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSPC7967-1G
PC7967-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 255 zł
255 zł
-
+
Add to cart
1 * 1 g
255 zł
-
+
Add to cart
APOSPC7967-5G
PC7967-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 899 zł
899 zł
-
+
Add to cart
1 * 5 g
899 zł
-
+
Add to cart