Order Entry
b.safe Waste cap electronic fill-level control
b.safe Waste cap electronic fill-level control
Supplier:  Bohlender
Certificates
undefined
b.safe Waste cap electronic fill-level control
Supplier:  Bohlender
Related Information
Product Information
  • Catalog #
    Supplier #
    Thread
    Pk
    Availability
    Price
  • Waste cap, electronic fill-level control, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    Waste cap, electronic fill-level control, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    590-0102
    M206-41
    S60
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 950 zł
    1 950 zł
    1 * 1 SZT
       1 950 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0103
    M206-42
    S60
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 900 zł
    1 900 zł
    1 * 1 SZT
       1 900 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control with grounding connection, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    Waste cap, electronic fill-level control with grounding connection, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    590-0105
    M206-60
    S60
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 030 zł
    2 030 zł
    1 * 1 SZT
       2 030 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control with grounding connection, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control with grounding connection, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0106
    M206-61
    S60
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 040 zł
    2 040 zł
    1 * 1 SZT
       2 040 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, filling funnel, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, filling funnel, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0104
    M206-51
    S60
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 960 zł
    1 960 zł
    1 * 1 SZT
       1 960 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    Waste cap, electronic fill-level control, 3× capillary connector, incl. 3× fittings for tubing Ext.Ø 1,6 mm, 3× fittings for tubing Ext.Ø 2,2 mm, 3× fittings for tubing Ext.Ø 3,2 mm, 2× blind fittings
    590-0100
    M205-40
    S55
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 870 zł
    1 870 zł
    1 * 1 SZT
       1 870 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fitting, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fitting, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0101
    M205-41
    S55
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 950 zł
    1 950 zł
    1 * 1 SZT
       1 950 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fitting
    Waste cap, electronic fill-level control, 2× capillary connector, incl. 2× fittings for tubing Ext.Ø 1,6 mm, 2× fittings for tubing Ext.Ø 2,2 mm, 2× fittings for tubing Ext.Ø 3,2 mm, 1× blind fitting
    590-0098
    M202-40
    S51
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 950 zł
    1 950 zł
    1 * 1 SZT
       1 950 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0109
    M210-41
    S95
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 350 zł
    2 350 zł
    1 * 1 SZT
       2 350 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0108
    M209-41
    S90
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 220 zł
    2 220 zł
    1 * 1 SZT
       2 220 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 1× capillary connector, incl. 1× fitting for tubing Ext.Ø 1,6 mm, 1× fitting for tubing Ext.Ø 2,2 mm, 1× fitting for tubing Ext.Ø 3,2 mm, 1× blind fitting, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 1× capillary connector, incl. 1× fitting for tubing Ext.Ø 1,6 mm, 1× fitting for tubing Ext.Ø 2,2 mm, 1× fitting for tubing Ext.Ø 3,2 mm, 1× blind fitting, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0099
    M203-41
    B53
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 370 zł
    1 370 zł
    1 * 1 SZT
       1 370 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    Waste cap, electronic fill-level control, 4× capillary connector, incl. 4× fittings for tubing Ext.Ø 1,6 mm, 4× fittings for tubing Ext.Ø 2,2 mm, 4× fittings for tubing Ext.Ø 3,2 mm, 3× blind fittings, 1× hose connector for tubing Int.Ø 5,5 - 8,0 mm
    590-0107
    M208-41
    B83
    1
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 2 510 zł
    2 510 zł
    1 * 1 SZT
       2 510 zł
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Exhaust filters
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Exhaust filter, medium, 6 months service life
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 710 zł
    710 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 03
    Exhaust filter, large, 12 months service life
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 905 zł
    905 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Exhaust filter, small, 3 months service life
    2
    Log in for availability
    Log in for availability
    1 * 2 SZT
    1 * 2 SZT - 425 zł
    425 zł
Replacement hose connectors
Product Information
  • Catalog #
    Seq
    Description
    Material
    Hole Ø
    Length
    Thread
    Tubing int.Ø
    Pk
    Availability
    Price
  • 06
    Hose connector, bent, yellow
    PP
    4,0 mm
    45 mm
    NPT ¹/₈"
    5,5 - 8,0 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 54,50 zł
    54,50 zł
Product Information
  • Catalog #
    Seq
    Description
    Material
    Hole Ø
    Length
    Thread
    Tubing int.Ø
    Pk
    Availability
    Price
  • 12
    Hose connector, straight, milky white
    PP
    7,0 mm
    40 mm
    NPT ¹/₈"
    9,0 - 10,5 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 45,50 zł
    45,50 zł
Adaptors for exhaust filter
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Adaptor for exhaust filter 0°
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 155 zł
    155 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Adaptor for exhaust filter 1× 45°
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 244 zł
    244 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 03
    Adaptor for exhaust filter 1× 90°
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 220 zł
    220 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 04
    Adaptor for exhaust filter 2× 90°
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 309 zł
    309 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 05
    Adaptor for capillary connection
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 13,50 zł
    13,50 zł
Adaptors for exhaust filter connection
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Exhaust filter connection UNF ¹/₄", PTFE
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 138 zł
    138 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Exhaust filter connection UNF ¹/₄", PTFE-EX
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 156 zł
    156 zł
Replacement fittings
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 02
    Fitting
    Lilac
    2,2 mm (⁷/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 159 zł
    159 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 03
    Fitting
    Grey
    2,2 mm (⁷/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 159 zł
    159 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 04
    Fitting
    White
    3,2 mm (¹/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 161 zł
    161 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 05
    Fitting
    Blue
    3,2 mm (¹/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 159 zł
    159 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 06
    Fitting
    Red
    3,2 mm (¹/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 161 zł
    161 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 07
    Fitting
    Yellow
    3,2 mm (¹/₈")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 158 zł
    158 zł
Product Information
  • Catalog #
    Seq
    Description
    Colour
    For tubing ext.Ø
    Pk
    Availability
    Price
  • 01
    Fitting
    Green
    1,6 mm (¹/₁₆")
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 159 zł
    159 zł
Replacement blind fittings
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 01
    Blind fitting, UNF ¹/₄"
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 185 zł
    185 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 02
    Blind fitting, GL14
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 113 zł
    113 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 03
    Blind fitting, NPT ¹/₈"
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 92,50 zł
    92,50 zł
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 04
    Blind fitting, NS 29/32
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 380 zł
    380 zł