Order Entry
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoyl chloride
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoyl chloride
Supplier:  Apollo Scientific
Certificates
3-Bromo-2,6-dichloro-5-(trifluoromethyl)benzoyl chloride
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSPC303505-1G
    PC303505-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 160 zł
    1 160 zł
    1 * 1 g
       1 160 zł
    Supplier Article Number: PC303505-1G
    Local Article Number: APOSPC303505-1G
  • APOSPC303505-5G
    PC303505-5G
    Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 3 370 zł
    3 370 zł
    1 * 5 g
       3 370 zł
    Supplier Article Number: PC303505-5G
    Local Article Number: APOSPC303505-5G