Drukuj
Podłoże PER.C6, HyClone™ CDM4Retino (produkty HyClone (Cytiva))
Dostawca: HyClone products (Cytiva)
HyClone™
HYCLSH30519.01EA 0 PLN
HYCLSH30519.01
Podłoże PER.C6, HyClone™ CDM4Retino (produkty HyClone (Cytiva))
Pożywki do hodowli komórkowych
HyClone™ CDM4Retino is a chemically-defined liquid medium containing no proteins, animal derived components, hydrolysates or other undefined components. This high performance cell culture medium is developed through Metabolic Pathway Design to increase the process yields for the industrial manufacture of adenoviral vectors and rproteins using PER.C6 cells. Manufactured according to cGMP guidelines.
  • Ready-to-use liquid formulation requires no supplementation
  • Allows for direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies