Order Entry
ICP-MS Nebulisers and Supplies for PerkinElmer
ICP-MS Nebulisers and Supplies for PerkinElmer
Supplier:  Agilent
Certificates
undefined
ICP-MS Nebulisers and Supplies for PerkinElmer
Supplier:  Agilent
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Clip for type A/C/K concentric nebulisers, for ELAN 9000/6xX00/DRC/NexION
    Clip for type A/C/K concentric nebulisers, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0480
    8003-0480
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 40,20 zł
    40,20 zł
    1 * 1 SZT
       40,20 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Clip, nebuliser pressure hose
    Clip, nebuliser pressure hose
    HEWL8003-0495
    8003-0495
    10
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 235 zł
    235 zł
    1 * 10 SZT
       235 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Cross-Flow II replacement GemTips, PEEK, orifices color-coded, argon, 0.23 mm (0.009") ruby red, sample, 0.34 mm (0.013") clear sapphire
    Cross-Flow II replacement GemTips, PEEK, orifices color-coded, argon, 0.23 mm (0.009") ruby red, sample, 0.34 mm (0.013") clear sapphire
    HEWL8003-0484
    8003-0484
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 550 zł
    1 550 zł
    1 * 1 SZT
       1 550 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • End cap, for Cross-Flow II nebuliser, for all GemClean Cross-Flow II nebulisers after September 1998, for ELAN 9000/6xX00/DRC/NexION
    End cap, for Cross-Flow II nebuliser, for all GemClean Cross-Flow II nebulisers after September 1998, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0487
    8003-0487
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 240 zł
    1 240 zł
    1 * 1 SZT
       1 240 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Ferrule kit, for Cross-Flow II nebuliser. Includes sample and argon ferrules, for all GemClean Cross-Flow II nebulisers after September 1998
    Ferrule kit, for Cross-Flow II nebuliser. Includes sample and argon ferrules, for all GemClean Cross-Flow II nebulisers after September 1998
    HEWL8003-0486
    8003-0486
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 999 zł
    999 zł
    1 * 1 SZT
       999 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Ferrule, argon, for Cross-Flow II nebuliser
    Ferrule, argon, for Cross-Flow II nebuliser
    HEWL8003-0485
    8003-0485
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 141 zł
    141 zł
    1 * 1 SZT
       141 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Ferrule, sample, for Cross-Flow II nebuliser, for all GemClean Cross-Flow II nebulisers after September 1998
    Ferrule, sample, for Cross-Flow II nebuliser, for all GemClean Cross-Flow II nebulisers after September 1998
    HEWL8003-0488
    8003-0488
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 168 zł
    168 zł
    1 * 1 SZT
       168 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Fitting, female barb, chlorotrifluoroethylene, for sample inlet fitting used with type A/C/K nebulisers
    Fitting, female barb, chlorotrifluoroethylene, for sample inlet fitting used with type A/C/K nebulisers
    HEWL8003-0503
    8003-0503
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 724 zł
    724 zł
    1 * 1 SZT
       724 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Fitting, male barb, chlorotrifluoroethylene, for sample inlet fitting used with type A/C/K nebulisers
    Fitting, male barb, chlorotrifluoroethylene, for sample inlet fitting used with type A/C/K nebulisers
    HEWL8003-0502
    8003-0502
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 724 zł
    724 zł
    1 * 1 SZT
       724 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, GemClean Cross-Flow II with end cap, GemTip clear sapphire tip, GemTip red ruby argon tip, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, GemClean Cross-Flow II with end cap, GemTip clear sapphire tip, GemTip red ruby argon tip, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0483
    8003-0483
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 910 zł
    6 910 zł
    1 * 1 SZT
       6 910 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, glass, concentric, type C3, argon flow 1 L/min, uptake 3 ml/min, for high TDS samples, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, glass, concentric, type C3, argon flow 1 L/min, uptake 3 ml/min, for high TDS samples, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0475
    8003-0475
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 590 zł
    7 590 zł
    1 * 1 SZT
       7 590 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, glass, concentric, type K3, argon flow 1 L/min, uptake 3 ml/min, for high TDS samples, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, glass, concentric, type K3, argon flow 1 L/min, uptake 3 ml/min, for high TDS samples, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0476
    8003-0476
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 130 zł
    6 130 zł
    1 * 1 SZT
       6 130 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, glass, type C0,5, argon flow 1 L/min, uptake 0,5 ml/min, for samples with high TDS, for NexION
    Nebuliser, glass, type C0,5, argon flow 1 L/min, uptake 0,5 ml/min, for samples with high TDS, for NexION
    HEWL8003-0479
    8003-0479
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 830 zł
    6 830 zł
    1 * 1 SZT
       6 830 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, MicroMist, micro-uptake, glass, concentric, uptake 0,2 ml/min, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, MicroMist, micro-uptake, glass, concentric, uptake 0,2 ml/min, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0589
    8003-0589
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 8 750 zł
    8 750 zł
    1 * 1 SZT
       8 750 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Union, chlorotrifluoroethylene, for PFA-ST Nebuliser, connects to ultraclean sampler probe
    Union, chlorotrifluoroethylene, for PFA-ST Nebuliser, connects to ultraclean sampler probe
    HEWL8003-0506
    8003-0506
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 959 zł
    959 zł
    1 * 1 SZT
       959 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, OpalMist PFA, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, OpalMist PFA, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0500
    8003-0500
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 13 380 zł
    13 380 zł
    1 * 1 SZT
       13 380 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, PFA-ST PTFE, ultraclean, with exchangeable capillaries to provide flow rates from 100 to 400 ul/min
    Nebuliser, PFA-ST PTFE, ultraclean, with exchangeable capillaries to provide flow rates from 100 to 400 ul/min
    HEWL8003-0505
    8003-0505
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 15 860 zł
    15 860 zł
    1 * 1 SZT
       15 860 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, PFA-ST3, PTFE, microflow, high performance, clog resistant, chemically inert, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, PFA-ST3, PTFE, microflow, high performance, clog resistant, chemically inert, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0507
    8003-0507
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 18 630 zł
    18 630 zł
    1 * 1 SZT
       18 630 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, quartz, type A0,5, high-sensitivity for ultratrace applications, argon flow 1 L/min, uptake 0,5 ml/min, for NexION
    Nebuliser, quartz, type A0,5, high-sensitivity for ultratrace applications, argon flow 1 L/min, uptake 0,5 ml/min, for NexION
    HEWL8003-0478
    8003-0478
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 560 zł
    6 560 zł
    1 * 1 SZT
       6 560 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, quartz, type A3, high-sensitivity for ultratrace applications, argon flow 1 L/min, uptake 3 ml/min, for ELAN 9000/6xX00/DRC
    Nebuliser, quartz, type A3, high-sensitivity for ultratrace applications, argon flow 1 L/min, uptake 3 ml/min, for ELAN 9000/6xX00/DRC
    HEWL8003-0477
    8003-0477
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 010 zł
    7 010 zł
    1 * 1 SZT
       7 010 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, U-series, MicroMist micro-uptake, glass, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, U-series, MicroMist micro-uptake, glass, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0489
    8003-0489
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 570 zł
    6 570 zł
    1 * 1 SZT
       6 570 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, U-series, SeaSpray, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, U-series, SeaSpray, concentric, uptake 0,4 ml/min, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0492
    8003-0492
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 8 230 zł
    8 230 zł
    1 * 1 SZT
       8 230 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Nebuliser, U-series, SeaSpray, concentric, uptake 2,0 ml/min, for ELAN 9000/6xX00/DRC/NexION
    Nebuliser, U-series, SeaSpray, concentric, uptake 2,0 ml/min, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0490
    8003-0490
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 8 190 zł
    8 190 zł
    1 * 1 SZT
       8 190 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Obstruction removal kit, for PFA nebulisers
    Obstruction removal kit, for PFA nebulisers
    HEWL8003-0508
    8003-0508
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 400 zł
    1 400 zł
    1 * 1 SZT
       1 400 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample capillary, PTFE, 0,508 mm Int.Ø, 1,57 mm Ext.Ø, for sample inlet fitting used with type A/C/K nebulisers
    Sample capillary, PTFE, 0,508 mm Int.Ø, 1,57 mm Ext.Ø, for sample inlet fitting used with type A/C/K nebulisers
    HEWL8003-0504
    8003-0504
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 101 zł
    101 zł
    1 * 1 SZT
       101 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample inlet (UniFit) fitting, 0.75 mm Int.Ø, 700 mm, for U-series concentric glass nebuliser
    Sample inlet (UniFit) fitting, 0.75 mm Int.Ø, 700 mm, for U-series concentric glass nebuliser
    HEWL8003-0493
    8003-0493
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 603 zł
    603 zł
    1 * 1 SZT
       603 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample inlet fitting for type A/C/K concentric nebulisers, for ELAN 9000/6xX00/DRC/NexION
    Sample inlet fitting for type A/C/K concentric nebulisers, for ELAN 9000/6xX00/DRC/NexION
    HEWL8003-0481
    8003-0481
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 851 zł
    851 zł
    1 * 1 SZT
       851 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample introduction kit, HF-resistant, for NexION
    Sample introduction kit, HF-resistant, for NexION
    HEWL8003-0513
    8003-0513
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 30 900 zł
    30 900 zł
    1 * 1 SZT
       30 900 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample introduction kit, HF-resistant, PFA-platinum, for analysis of semiconductor-grade chemicals and Si wafers, for NexION
    Sample introduction kit, HF-resistant, PFA-platinum, for analysis of semiconductor-grade chemicals and Si wafers, for NexION
    HEWL8003-0511
    8003-0511
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 104 460 zł
    104 460 zł
    1 * 1 SZT
       104 460 zł
    Specifications:
    Specifications
    Environmentally Preferable
  • Sample introduction kit, HF-resistant, PFA-Sapphire, chemically inert and clean introduction system, for NexION
    Sample introduction kit, HF-resistant, PFA-Sapphire, chemically inert and clean introduction system, for NexION
    HEWL8003-0512
    8003-0512
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 59 980 zł
    59 980 zł
    1 * 1 SZT
       59 980 zł
    Specifications:
    Specifications
    Environmentally Preferable