Drukuj
PC thin membrane, Whatman™ Cyclopore™
Dostawca: Whatman products (Cytiva)
Whatman™
514-0948EA 760 PLN
514-0948
PC thin membrane, Whatman™ Cyclopore™
Filtry Filtry membranowe
Cytiva's Whatman Cyclopore polycarbonate membranes are true pore size microporous membranes featuring the sharp cut-off and reproducible microfiltration characteristics of track-etched membrane filters.
  • Low affinity for stains improves visibility under a microscope
  • Low protein binding minimizes sample loss of proteinaceous samples
  • Narrow pore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
Cytiva's business offers a variety of Whatman PCTE membrane filters for gravimetric analyses. Browse the range of Whatman track-etched membranes to find other Cyclopore membranes and Nuclepore application-specific membranes for organic halide adsorption and electron microscopy.
Whatman Cyclopore track-etched membrane filters are appropriate for a wide variety of analytical procedures such as densitometry, gravimetric analysis, and emission spectroscopy.