Drukuj
Zestaw do usówania rozlanych chemikaliów w mobilnym pojemniku w żółtym kolorze o wysokiej widoczności, PIG® HazMat
Dostawca: New Pig
PIG® HazMat
NEWPKIT366EA 0 PLN
NEWPKIT366
Zestaw do usówania rozlanych chemikaliów w mobilnym pojemniku w żółtym kolorze o wysokiej widoczności, PIG® HazMat
Materiały do kontroli wycieków
Bright-yellow 50-gallon container resists chemicals and stands out so you can locate it quickly.
  • Hinged lid provides easy access
  • Specially treated for unsurpassed performance
  • Heavy-duty 203 mm rubber wheels roll easily over nearly any surface
  • Resist chemicals
Lightweight, 100% polyethylene container resists chemicals. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulphuric acid and 30% sodium hydroxide. PIG HazMat Socks stop spreading spills; PIG HazMat Pads absorb quickly.