Order Entry
Large laboratory glasswashers, PLW 8615
Large laboratory glasswashers, PLW 8615
Supplier:  MIELE
Certificates
undefined
Large laboratory glasswashers, PLW 8615
Supplier:  MIELE
Specifications
  • Catalog No:
  • MIEL11176250
  • MIEL11176260
  • MIEL11176280
  • MIEL11176270
  • Model:
  • PLW 8615
  • PLW 8615
  • PLW 8615
  • PLW 8615
  • Dispensers:
  • 2 dispenser for liquid chemicals;
    4 dispenser connections extra available
  • 2 dispenser for liquid chemicals;
    4 dispenser connections extra available
  • 2 dispenser for liquid chemicals;
    4 dispenser connections extra available
  • 2 dispenser for liquid chemicals;
    4 dispenser connections extra available
  • Drainage:
  • 1 drain valve connection (unless it is drain pump version)
  • 1 drain valve connection (unless it is drain pump version)
  • 1 drain valve connection (unless it is drain pump version)
  • 1 drain valve connection (unless it is drain pump version)
  • Electrical:
  • 3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
  • 3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
  • 3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
  • 3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
  • Plumbing:
  • 2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
  • 2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
  • 2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
  • 2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
  • W×D×H:
  • 2495×900×990 mm
  • 2495×900×990 mm
  • 2495×900×990 mm
  • 2495×900×990 mm
  • Weight:
  • 550 kg
  • 550 kg
  • 550 kg
  • 550 kg
Specifications
Large laboratory glasswashers
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Large laboratory glasswasher PLW 8615 EL, electrically heated - single door, 900 mm wide, 351 L usable capacity
    Large laboratory glasswasher PLW 8615 EL, electrically heated - single door, 900 mm wide, 351 L usable capacity
    MIEL11176250
    11176250
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 11176250
    Local Article Number: MIEL11176250
    Specifications:
    More
    Accessories
    Specifications
    Model
    PLW 8615
    Environmentally Preferable
    Dispensers
    2 dispenser for liquid chemicals;
    4 dispenser connections extra available
    Drainage
    1 drain valve connection (unless it is drain pump version)
    Electrical
    3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
    Plumbing
    2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
    W×D×H
    2495×900×990 mm
    Weight
    550 kg
  • Large laboratory glasswasher PLW 8615 EL CT, electrically heated - single door, with combi tank, 900 mm wide, 351 L usable capacity
    Large laboratory glasswasher PLW 8615 EL CT, electrically heated - single door, with combi tank, 900 mm wide, 351 L usable capacity
    MIEL11176260
    11176260
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 11176260
    Local Article Number: MIEL11176260
    Specifications:
    More
    Accessories
    Specifications
    Model
    PLW 8615
    Environmentally Preferable
    Dispensers
    2 dispenser for liquid chemicals;
    4 dispenser connections extra available
    Drainage
    1 drain valve connection (unless it is drain pump version)
    Electrical
    3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
    Plumbing
    2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
    W×D×H
    2495×900×990 mm
    Weight
    550 kg
  • Large laboratory glasswasher PLW 8615 EL/S, steam/electrically heated - single door, 900 mm wide, 351 L usable capacity
    Large laboratory glasswasher PLW 8615 EL/S, steam/electrically heated - single door, 900 mm wide, 351 L usable capacity
    MIEL11176270
    11176270
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 11176270
    Local Article Number: MIEL11176270
    Specifications:
    More
    Accessories
    Specifications
    Model
    PLW 8615
    Environmentally Preferable
    Dispensers
    2 dispenser for liquid chemicals;
    4 dispenser connections extra available
    Drainage
    1 drain valve connection (unless it is drain pump version)
    Electrical
    3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
    Plumbing
    2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
    W×D×H
    2495×900×990 mm
    Weight
    550 kg
  • Large laboratory glasswasher PLW 8615 EL/S CT, steam/electrically heated - single door, with combi tank, 900 mm wide, 351 L usable capacity
    Large laboratory glasswasher PLW 8615 EL/S CT, steam/electrically heated - single door, with combi tank, 900 mm wide, 351 L usable capacity
    MIEL11176280
    11176280
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT
    Supplier Article Number: 11176280
    Local Article Number: MIEL11176280
    Specifications:
    More
    Accessories
    Specifications
    Model
    PLW 8615
    Environmentally Preferable
    Dispensers
    2 dispenser for liquid chemicals;
    4 dispenser connections extra available
    Drainage
    1 drain valve connection (unless it is drain pump version)
    Electrical
    3 AC, 208 V, 60 Hz;
    Fuse 32 - 35 A
    Plumbing
    2× cold water (CW);
    1× hotwater (HW)
    2× de-mineralised water (AD)
    W×D×H
    2495×900×990 mm
    Weight
    550 kg
Components
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 53
    DU 900 Drying unit (900 mm)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    Supplier Article Number: 10880710
    Local Article Number: MIEL10880710
Product Information
  • Catalog #
    Seq
    Description
    Pk
    Availability
    Price
  • 56
    RP 900 Rolling Plinth (900 mm)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    Supplier Article Number: 10941940
    Local Article Number: MIEL10941940
Delivery and installation
Product Information
  • Catalog #
    Description
    Pk
    Availability
    Price
  • Delivery and installation service for Miele glasswashers
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    Supplier Article Number: 10506510
    Local Article Number: MIEL10506510