Order Entry
Standard use piggyback labels
Standard use piggyback labels
Supplier:  GA
Certificates
undefined
Standard use piggyback labels
Supplier:  GA
Product Information
  • Catalog #
    Supplier #
    Printing method
    Colour
    Printer model
    Pk
    Availability
    Price
  • Standard piggyback labels, core size 1"
    Standard piggyback labels, core size 1"
    GAINPID-2C1-0.5WH
    PID-2C1-0.5WH
    White
    1 Roll
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 282 zł
    282 zł
    1 * 1 Roll
       282 zł
    Supplier Article Number: PID-2C1-0.5WH
    Local Article Number: GAINPID-2C1-0.5WH
    Specifications:
    Specifications
    Environmentally Preferable
  • Standard piggyback labels, core size 3"
    Standard piggyback labels, core size 3"
    GAINPID-2C3-1WH
    PID-2C3-1WH
    White
    1 Roll
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 565 zł
    565 zł
    1 * 1 Roll
       565 zł
    Supplier Article Number: PID-2C3-1WH
    Local Article Number: GAINPID-2C3-1WH
    Specifications:
    Specifications
    Environmentally Preferable