Drukuj
Native Barcoding Expansion (Q20+)
Dostawca: Oxford Nanopore Technologies
OXNTEXP-NBD112EA 1710 PLN
OXNTEXP-NBD112
Native Barcoding Expansion (Q20+)
Odczynniki do kwasów nukleinowych Odczynniki do sekwencjonowania nowej generacji
Additional library preparation reagents for Native Barcoding Kit 24 and 96 (SQK-NBD112.24 and SQK-NBD112.96).
This kit contains additional library preparation reagents for the Native Barcoding Kit 24 and 96 (SQK-NBD112.24 and SQK-NBD112.96) for another 12 reactions. Additional reagents include Adapter Mix II H (AMII H), AMPure XP Beads (AXP), EDTA (EDTA) and L Fragment Buffer (LFB). Flow cell priming reagents are also included; Flush Buffer (FB) and Flush Tether (FLT).
For sample multiplexing see Native Barcoding Kit 24 and Native Barcoding Kit 96.
Dodatkowe informacje: Flow cells and kits are shipped together at 2 to 8°C. Upon receipt, please place the product in a long-term storage location at -20°C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Uwaga: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.