Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Ideal for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). Just slip your flexible tubing or hose onto the barb to create a secure seal. 316 stainless steel material features very good chemical resistance with a wide temperature range. Sterilise via ethylene oxide, autoclave, or gamma irradiation.
- Environmentally Preferable:
- Model:
- 30620-11
- 30620-12
- 30620-09
- 31208-14
- 30620-10
- 31208-15
- 31208-13
- Orifice dimensions:
- 4,763 mm (0,1875")
- 8,73 mm (0,34375")
- —
- 0,7925 mm (0,03125")
- —
- 4,763 mm (0,1875")
- 0,7925 mm (0,03125")
- Sterilisation:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- —
- Autoclave, ethylene oxide (ETO), or gamma
- —
- —
- Warranty:
- —
- 1 Month
- 1 Month
- 1 year
- 1 Month
- 1 year
- 1 year
- Barb size/tubing int.Ø (port 1)Barb size/tubing int.Ø (port 2)Connection TypeFitting materialFlow patternPk
- Barbed fittings, reducing connector, stainless steel, tubing ID: ½×¼"Barbed fittings, reducing connector, stainless steel, tubing ID: ½×¼"½"¼"Barbed reducerStainless steel 316Straight11 * 1 SZT374 zł: MFLA30620-11
Specifications
Model30620-11Environmentally PreferableOrifice dimensions4,763 mm (0,1875")SterilisationAutoclave, ethylene oxide (ETO), or gammaWarranty— - Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¼×³⁄₁₆"Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¼×³⁄₁₆"¼"³⁄₁₆"Barbed reducerStainless steel 316Straight11 * 1 SZT210 zł: MFLA30620-10
Specifications
Model30620-10Environmentally PreferableOrifice dimensions—SterilisationAutoclave, ethylene oxide (ETO), or gammaWarranty1 Month - Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¼×⅛"Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¼×⅛"¼"⅛"Barbed reducerStainless steel 316Straight11 * 1 SZT382 zł: MFLA31208-14
Specifications
Model31208-14Environmentally PreferableOrifice dimensions0,7925 mm (0,03125")Sterilisation—Warranty1 year - Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¾×½"Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ¾×½"¾"½"Barbed reducerStainless steel 316Straight11 * 1 SZT380 zł: MFLA30620-12
Specifications
Model30620-12Environmentally PreferableOrifice dimensions8,73 mm (0,34375")SterilisationAutoclave, ethylene oxide (ETO), or gammaWarranty1 Month - Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ³⁄₁₆×⅛"Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ³⁄₁₆×⅛"³⁄₁₆"³⁄₁₆"Barbed reducerStainless steel 316Straight11 * 1 SZT424 zł: MFLA31208-13
Specifications
Model31208-13Environmentally PreferableOrifice dimensions0,7925 mm (0,03125")Sterilisation—Warranty1 year - Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ⅜×¼"Fitting, 316 stainless steel, straight, hose barb reducer, tubing ID: ⅜×¼"⅜"¼"Barbed reducerStainless steel 316Straight11 * 1 SZT396 zł: MFLA31208-15
Specifications
Model31208-15Environmentally PreferableOrifice dimensions4,763 mm (0,1875")Sterilisation—Warranty1 year - Fitting, 316 stainless steel, straight, reduction coupler, tubing ID: ⅛׳⁄₃₂"Fitting, 316 stainless steel, straight, reduction coupler, tubing ID: ⅛׳⁄₃₂"⅛"³⁄₃₂"Barbed reducerStainless steel 316Straight11 * 1 SZT142 zł: MFLA30620-09
Specifications
Model30620-09Environmentally PreferableOrifice dimensions—SterilisationAutoclave, ethylene oxide (ETO), or gammaWarranty1 Month