Order Entry
Micropump® A-Mount Cavity-Style Pump Heads
Micropump® A-Mount Cavity-Style Pump Heads
Certificates
undefined
Micropump® A-Mount Cavity-Style Pump Heads
Specifications
  • Body material:
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • Differential pressure:
  • 5,5 bar (80 psi)
  • 5,5 bar (80 psi)
  • 5,5 bar (80 psi)
  • 3,4 bar (50 psi)
  • 3,4 bar (50 psi)
  • 3,4 bar (50 psi)
  • 4,8 bar (70 psi)
  • 5,2 bar (75 psi)
  • 4,8 bar (70 psi)
  • Duty cycle continuous:
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Yes, 150 to 5000 rpm
  • Gear material:
  • PEEK
  • PEEK
  • PPS
  • PEEK
  • PTFE
  • PTFE
  • PTFE
  • PEEK
  • PPS
  • PEEK
  • Inlet connection:
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 1/8" NPT
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • Internal bypass:
  • No
  • Yes
  • No
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • No
  • Max. temperature:
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • 99 °C (210 °F)
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • 54 °C (130 °F)
  • Max. viscosity:
  • 1500 cP
  • 1500 cP
  • 1500 cP
  • 100 cP
  • 1500 cP
  • 1500 cP
  • 1500 cP
  • 1500 cP
  • 1500 cP
  • Min. temperature:
  • –43 °C (–45 °F)
  • –43 °C (–45 °F)
  • –43 °C (–45 °F)
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • –43 °C (–45 °F)
  • –46 °C (–50 °F)
  • –43 °C (–45 °F)
  • Min. viscosity:
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • 0,2 cP
  • Outlet connection:
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 1/8" NPT
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • Particulates:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Relief valve:
  • No
  • Yes
  • No
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • No
  • Reversible:
  • +
  • -
  • +
  • -
  • -
  • -
  • -
  • -
  • +
  • Run dry:
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Seal material:
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • Series:
  • GJ-N23
  • GJ-N21
  • GJ-N23
  • GJ-N21
  • GJ-N23
  • GJ-N21
  • GJ-N25
  • GJ-N25
  • GJ-N25
  • Suction lift, wet:
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • 610 mm of H₂O (24" H₂O)
  • Wetted materials:
  • 316 SS, PEEK, PTFE
  • 316 SS, PEEK, PTFE
  • 316 SS, PEEK, PTFE
  • 316 SS, PTFE
  • 316 SS, PTFE
  • 316 SS, PTFE
  • 316 SS, PEEK, PTFE
  • 316 SS, PPS, PTFE
  • 316 SS, PEEK, PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Information
  • Catalog #
    Flow rate
    Maximum Flow Rate
    Max. motor RPM
    Pk
    Availability
    Price
  • A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,64 ml/rev
    A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,64 ml/rev
    MFLX73011-08
    0,64 ml/rev
    3200 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 400 zł
    7 400 zł
    1 * 1 SZT
       7 400 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    5,5 bar (80 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PEEK
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    No
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –43 °C (–45 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    No
    Reversible
    +
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N23
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PEEK, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; Bypass/SS/PEEK/PTFE; 0,32 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/SS/PEEK/PTFE; 0,32 ml/rev
    MFLX73011-02
    0,32 ml/rev
    1600 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 720 zł
    6 720 zł
    1 * 1 SZT
       6 720 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    5,5 bar (80 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PEEK
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –43 °C (–45 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N21
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PEEK, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; SS/PPS/PTFE; 0,64 ml/rev
    A-Mount Cavity Style Pump Head; SS/PPS/PTFE; 0,64 ml/rev
    MFLX74012-21
    0,64 ml/Umdrehung
    3200 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 350 zł
    7 350 zł
    1 * 1 SZT
       7 350 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    Differential pressure
    Duty cycle continuous
    Gear material
    PPS
    Inlet connection
    1/8" NPT
    Internal bypass
    Max. temperature
    Max. viscosity
    Min. temperature
    Min. viscosity
    Outlet connection
    1/8" NPT
    Particulates
    Relief valve
    Reversible
    Run dry
    Seal material
    Series
    GJ-N23
    Suction lift, wet
    Wetted materials
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,32 ml/rev
    A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,32 ml/rev
    MFLX73011-00
    0,32 ml/rev
    1600 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 830 zł
    7 830 zł
    1 * 1 SZT
       7 830 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    5,5 bar (80 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PEEK
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    No
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –43 °C (–45 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    No
    Reversible
    +
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N21
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PEEK, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,64 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,64 ml/rev
    MFLX07002-23
    0,64 ml/rev
    3200 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 300 zł
    7 300 zł
    1 * 1 SZT
       7 300 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    3,4 bar (50 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PTFE
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    100 cP
    Min. temperature
    –46 °C (–50 °F)
    Min. viscosity
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N23
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,32 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,32 ml/rev
    MFLX07002-17
    0,32 ml/rev
    1600 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 000 zł
    6 000 zł
    1 * 1 SZT
       6 000 zł
    Specifications:
    CSA Certified
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    3,4 bar (50 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PTFE
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –46 °C (–50 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N21
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PTFE
    Warranty
    1 year
    CSA Certified
  • A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,91 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/SS/PTFE/PTFE; 0,91 ml/rev
    MFLX07001-40
    0,91 ml/rev
    4550 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 010 zł
    7 010 zł
    1 * 1 SZT
       7 010 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    3,4 bar (50 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PTFE
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    99 °C (210 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –46 °C (–50 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N25
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; Bypass/SS/PEEK/PTFE; 0,91 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/SS/PEEK/PTFE; 0,91 ml/rev
    MFLX73011-18
    0,91 ml/rev
    4550 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 390 zł
    6 390 zł
    1 * 1 SZT
       6 390 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    4,8 bar (70 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PEEK
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –43 °C (–45 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N25
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PEEK, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; Bypass/PPS/PPS/PTFE; 0,64 ml/rev
    A-Mount Cavity Style Pump Head; Bypass/PPS/PPS/PTFE; 0,64 ml/rev
    MFLX07001-80
    0,64 ml/rev
    1600 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 7 640 zł
    7 640 zł
    1 * 1 SZT
       7 640 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    5,2 bar (75 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PPS
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    Yes
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –46 °C (–50 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    Yes
    Reversible
    -
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PPS, PTFE
    Warranty
    1 year
  • A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,91 ml/rev
    A-Mount Cavity Style Pump Head; SS/PEEK/PTFE; 0,91 ml/rev
    MFLX73011-16
    0,91 ml/rev
    4550 ml/min at 5000 rpm
    10000 rpm
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 6 630 zł
    6 630 zł
    1 * 1 SZT
       6 630 zł
    Specifications:
    More
    Accessories
    Specifications
    Environmentally Preferable
    Body material
    316 stainless steel
    Differential pressure
    4,8 bar (70 psi)
    Duty cycle continuous
    Yes, 150 to 5000 rpm
    Gear material
    PEEK
    Inlet connection
    0,125 NPT(F)
    Internal bypass
    No
    Max. temperature
    54 °C (130 °F)
    Max. viscosity
    1500 cP
    Min. temperature
    –43 °C (–45 °F)
    Min. viscosity
    0,2 cP
    Outlet connection
    0,125 NPT(F)
    Particulates
    No
    Relief valve
    No
    Reversible
    +
    Run dry
    Under 30 seconds
    Seal material
    PTFE
    Series
    GJ-N25
    Suction lift, wet
    610 mm of H₂O (24" H₂O)
    Wetted materials
    316 SS, PEEK, PTFE
    Warranty
    1 year