Parker Hannifin Fitting, Black Polypropylene, Elbow, Compression Union, 3/8" OD
MFLX06444-42
- Connection Type:Compression Union
- Flow pattern:Elbow
- Description:Fitting, Black Polypropylene, Elbow, Compression Union, ⅜" OD
- Environmentally Preferable:
- Orifice dimensions:6,35 mm (0,25")
- Sterilisation:Autoclave only
- Pk:25
- Fitting material:Polypropylene
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
- Connection Type:Compression Union
- Flow pattern:Elbow
- Description:Fitting, Black Polypropylene, Elbow, Compression Union, ⅜" OD
- Environmentally Preferable:
- Orifice dimensions:6,35 mm (0,25")
- Sterilisation:Autoclave only
- Pk:25
- Fitting material:Polypropylene