Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
As flexible as you need it.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications.
Characteristics:Nontoxic DEHP-free formulation. Flexible, small bend radii. Transparent; allows for visual inspection and flow monitoring.
Certifications: FDA compliant, Proposition 65, REACH
Temperature range: -58 to 165 °F (-50 to 73 °C)
Sterilize: Autoclave.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX95505-44
- MFLX95505-26
- MFLX95505-28
- MFLX95505-42
- MFLX95505-00
- MFLX95505-02
- MFLX95505-04
- MFLX95505-06
- MFLX95505-14
- MFLX95505-18
- MFLX95505-20
- MFLX95505-22
- MFLX95505-24
- MFLX95505-16
- MFLX95505-08
- Model:
- 95505-44
- 95505-26
- 95505-28
- 95505-42
- 95505-00
- 95505-02
- 95505-04
- 95505-06
- 95505-14
- 95505-18
- 95505-20
- 95505-22
- 95505-24
- 95505-16
- 95505-08
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- Max. pressure:
- 1,24 bar (18 psi)
- 1,24 bar (18 psi)
- 1,72 bar (25 psi)
- 1,45 bar (21 psi)
- 3,38 bar (49 psi)
- 2,2 bar (32 psi)
- 2,14 bar (31 psi)
- 3 bar (43 psi)
- 1,72 bar (25 psi)
- 3 bar (43 psi)
- 1,45 bar (21 psi)
- 1,93 bar (28 psi)
- 2,48 bar (36 psi)
- 2,34 bar (34 psi)
- 2,14 bar (31 psi)
- Max. temperature:
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- Min. temperature:
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- –50 °C (–58 °F)
- Regulatory approvals:
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- FDA;Non-DEHP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Working pressure:
- 18 psi
- 18 psi
- 25 psi
- 21 psi
- 49 psi
- 32 psi
- 31 psi
- 43 psi
- 25 psi
- 43 psi
- 21 psi
- 28 psi
- 36 psi
- 34 psi
- 31 psi
Specifications
Product Information
- Catalog #Supplier #Tubing int.ØWall thicknessTubing ext.ØTubing materialLengthAvailabilityPrice
- Transfer Tubing, Flexible PVC, 1/4" ID x 1/2" OD; 50 FtTransfer Tubing, Flexible PVC, 1/4" ID x 1/2" OD; 50 Ft95505-186,4 mm (0,25")3,2 mm12,8 mm (0,5")PVC15,2 mLog in for availability1 * 50 Feet1 850 złSupplier Article Number: 95505-18Local Article Number: MFLB95505-18Specifications:
More
Accessories
Specifications
Model95505-18Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure3 bar (43 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure43 psi - Transfer Tubing, Flexible PVC, 1/4" ID x 3/8" OD; 50 FtTransfer Tubing, Flexible PVC, 1/4" ID x 3/8" OD; 50 Ft95505-146,4 mm (0,25")1,6 mm9,6 mm (0,375")PVC15,2 mLog in for availability1 * 50 Feet1 200 złSupplier Article Number: 95505-14Local Article Number: MFLB95505-14Specifications:
More
Accessories
Specifications
Model95505-14Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,72 bar (25 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure25 psi - Transfer Tubing, Flexible PVC, 1/4" ID x 7/16" OD; 50 FtTransfer Tubing, Flexible PVC, 1/4" ID x 7/16" OD; 50 Ft95505-166,4 mm (0,25")2,4 mm11,2 mm (0,4375")PVC15,2 mLog in for availability1 * 50 Feet1 490 złSupplier Article Number: 95505-16Local Article Number: MFLB95505-16Specifications:
More
Accessories
Specifications
Model95505-16Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure2,34 bar (34 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure34 psi - Transfer Tubing, Flexible PVC, 1/8" ID x 1/4" OD; 50 FtTransfer Tubing, Flexible PVC, 1/8" ID x 1/4" OD; 50 Ft95505-063,2 mm (0,125")1,6 mm6,4 mm (0,25")PVC15,2 mLog in for availability1 * 50 Feet567 złSupplier Article Number: 95505-06Local Article Number: MFLB95505-06Specifications:
More
Accessories
Specifications
Model95505-06Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure3 bar (43 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure43 psi - Transfer Tubing, Flexible PVC, 1/8" ID x 3/16" OD; 50 FtTransfer Tubing, Flexible PVC, 1/8" ID x 3/16" OD; 50 Ft95505-043,2 mm (0,125")1,6 mm4,8 mm (0,1875")PVC15,2 mLog in for availability1 * 50 Feet293 złSupplier Article Number: 95505-04Local Article Number: MFLB95505-04Specifications:
More
Accessories
Specifications
Model95505-04Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure2,14 bar (31 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure31 psi - Transfer Tubing, Flexible PVC, 1/16" ID x 1/8" OD; 50 FtTransfer Tubing, Flexible PVC, 1/16" ID x 1/8" OD; 50 Ft95505-001,6 mm (0,0625")0,8 mm3,2 mm (0,125")PVC15,2 mLog in for availability1 * 50 Feet173 złSupplier Article Number: 95505-00Local Article Number: MFLB95505-00Specifications:
More
Accessories
Specifications
Model95505-00Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure3,38 bar (49 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure49 psi - Transfer Tubing, Flexible PVC, 3/4" ID x 1" OD; 50 FtTransfer Tubing, Flexible PVC, 3/4" ID x 1" OD; 50 Ft95505-4419,1 mm (0,75")3,2 mm25,1 mm (1")PVC15,2 mLog in for availability1 * 50 Feet2 870 złSupplier Article Number: 95505-44Local Article Number: MFLB95505-44Specifications:
More
Accessories
Specifications
Model95505-44Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,24 bar (18 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure18 psi - Transfer Tubing, Flexible PVC, 3/8" ID x 1/2" OD; 50 FtTransfer Tubing, Flexible PVC, 3/8" ID x 1/2" OD; 50 Ft95505-269,6 mm (0,375")1,6 mm12,8 mm (0,5")PVC15,2 mLog in for availability1 * 50 Feet1 100 złSupplier Article Number: 95505-26Local Article Number: MFLB95505-26Specifications:
More
Accessories
Specifications
Model95505-26Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,24 bar (18 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure18 psi - Transfer Tubing, Flexible PVC, 3/8" ID x 9/16" OD; 50 FtTransfer Tubing, Flexible PVC, 3/8" ID x 9/16" OD; 50 Ft95505-289,6 mm (0,375")2,4 mm14,4 mm (0,5625")PVC15,2 mLog in for availability1 * 50 Feet1 170 złSupplier Article Number: 95505-28Local Article Number: MFLB95505-28Specifications:
More
Accessories
Specifications
Model95505-28Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,72 bar (25 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure25 psi - Transfer Tubing, Flexible PVC, 3/16" ID x 5/16" OD; 50 FtTransfer Tubing, Flexible PVC, 3/16" ID x 5/16" OD; 50 Ft95505-084,8 mm (0,1875")1,6 mm7,9 mm (0,3125")PVC15,2 mLog in for availability1 * 50 Feet653 złSupplier Article Number: 95505-08Local Article Number: MFLB95505-08Specifications:
More
Accessories
Specifications
Model95505-08Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure2,14 bar (31 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure31 psi - Transfer Tubing, Flexible PVC, 3/32" ID x 5/32" OD; 50 FtTransfer Tubing, Flexible PVC, 3/32" ID x 5/32" OD; 50 Ft95505-022,4 mm (0,09375")0,8 mm4 mm (0,1562")PVC15,2 mLog in for availability1 * 50 Feet226 złSupplier Article Number: 95505-02Local Article Number: MFLB95505-02Specifications:
More
Accessories
Specifications
Model95505-02Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure2,2 bar (32 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure32 psi - Transfer Tubing, Flexible PVC, 5/8" ID x 7/8" OD; 50 FtTransfer Tubing, Flexible PVC, 5/8" ID x 7/8" OD; 50 Ft95505-4216 mm (0,625")3,2 mm22,4 mm (0,875")PVC15,2 mLog in for availability1 * 50 Feet2 510 złSupplier Article Number: 95505-42Local Article Number: MFLB95505-42Specifications:
More
Accessories
Specifications
Model95505-42Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,45 bar (21 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure21 psi - Transfer Tubing, Flexible PVC, 5/16" ID x 1/2" OD; 50 FtTransfer Tubing, Flexible PVC, 5/16" ID x 1/2" OD; 50 Ft95505-228 mm (0,3125")2,4 mm12,8 mm (0,5")PVC15,2 mLog in for availability1 * 50 Feet1 300 złSupplier Article Number: 95505-22Local Article Number: MFLB95505-22Specifications:
More
Accessories
Specifications
Model95505-22Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,93 bar (28 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure28 psi - Transfer Tubing, Flexible PVC, 5/16" ID x 7/16" OD; 50 FtTransfer Tubing, Flexible PVC, 5/16" ID x 7/16" OD; 50 Ft95505-208 mm (0,3125")1,6 mm11,2 mm (0,4375")PVC15,2 mLog in for availability1 * 50 Feet1 080 złSupplier Article Number: 95505-20Local Article Number: MFLB95505-20Specifications:
More
Accessories
Specifications
Model95505-20Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure1,45 bar (21 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure21 psi - Transfer Tubing, Flexible PVC, 5/16" ID x 9/16" OD; 50 FtTransfer Tubing, Flexible PVC, 5/16" ID x 9/16" OD; 50 Ft95505-248 mm (0,3125")3,2 mm14,4 mm (0,5625")PVC15,2 mLog in for availability1 * 50 Feet1 610 złSupplier Article Number: 95505-24Local Article Number: MFLB95505-24Specifications:
More
Accessories
Specifications
Model95505-24Environmentally PreferableClarityTranslucentLength per pack15,2 m (50 ft.)Max. pressure2,48 bar (36 psi)Max. temperature73 °C (165 °F)Min. temperature–50 °C (–58 °F)Regulatory approvalsFDA;Non-DEHPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoWorking pressure36 psi