Order Entry
Hand-Operated Chemical-Resistant Drum Pumps
Hand-Operated Chemical-Resistant Drum Pumps
Certificates
undefined
Hand-Operated Chemical-Resistant Drum Pumps
Specifications
  • Outlet connection:
  • Discharge tubing
  • Spigot
  • Spigot
  • Discharge tubing
  • Tube ext.Ø, inlet:
  • 3,2 cm (1,25")
  • 3,2 cm (1,25")
  • 3,2 cm (1,25")
  • 3,2 cm (1,25")
  • Tube length, inlet:
  • 60 cm (23,622")
  • 60 cm (23,622")
  • 95 cm (37,4")
  • 95 cm (37,4")
  • Tube length, outlet:
  • 120 cm (47,25")
  • 120 cm (47,25")
  • Wetted materials:
  • PTFE/FEP
  • PTFE/FEP
  • PTFE/FEP
  • PTFE/FEP
  • Warranty:
  • 15 Months
  • 15 Months
  • 15 Months
  • 15 Months
Specifications
Product Information
  • Catalog #
    Supplier #
    Flow rate
    Pk
    Availability
    Price
  • Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
    Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
    MFLX06043-97
    06043-97
    3,7 strokes/L (14 strokes/gal.)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 080 zł
    4 080 zł
    1 * 1 SZT
       4 080 zł
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Outlet connection
    Discharge tubing
    Tube ext.Ø, inlet
    3,2 cm (1,25")
    Tube length, inlet
    60 cm (23,622")
    Tube length, outlet
    120 cm (47,25")
    Wetted materials
    PTFE/FEP
    Warranty
    15 Months
  • Hand Operated Drum Pump with Spigot, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
    Hand Operated Drum Pump with Spigot, PTFE/FEP, 60 cm inlet tube, 270 mL/stroke
    MFLX06043-96
    06043-96
    3,7 strokes/L (14 strokes/gal.)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 3 960 zł
    3 960 zł
    1 * 1 SZT
       3 960 zł
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Outlet connection
    Spigot
    Tube ext.Ø, inlet
    3,2 cm (1,25")
    Tube length, inlet
    60 cm (23,622")
    Tube length, outlet
    Wetted materials
    PTFE/FEP
    Warranty
    15 Months
  • Hand Operated Drum Pump with Spigot, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke
    Hand Operated Drum Pump with Spigot, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke
    MFLX06043-98
    06043-98
    2,5 strokes/L (9,5 strokes/gal.)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 180 zł
    4 180 zł
    1 * 1 SZT
       4 180 zł
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Outlet connection
    Spigot
    Tube ext.Ø, inlet
    3,2 cm (1,25")
    Tube length, inlet
    95 cm (37,4")
    Tube length, outlet
    Wetted materials
    PTFE/FEP
    Warranty
    15 Months
  • Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke
    Hand Operated Drum Pump with Discharge Tubing, PTFE/FEP, 95 cm inlet tube, 400 mL/stroke
    MFLX06043-99
    06043-99
    2,5 strokes/L (9,5 strokes/gal.)
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 4 050 zł
    4 050 zł
    1 * 1 SZT
       4 050 zł
    Specifications:
    More
    Specifications
    Environmentally Preferable
    Outlet connection
    Discharge tubing
    Tube ext.Ø, inlet
    3,2 cm (1,25")
    Tube length, inlet
    95 cm (37,4")
    Tube length, outlet
    120 cm (47,25")
    Wetted materials
    PTFE/FEP
    Warranty
    15 Months