Foot-Operated Pump Dispensers for Barrels, Canisters, and Tanks
Supplier: Avantor Fluid Handling
Certificates
About this item
Keep hands-free for safe dispensing.
- Large-volume foot-operated bellows for high pumping capacity
- Universal bung constructed of soft PVC for 1-3/16" to 2-3/4" (30 to 70 mm) openings
- Telescopic immersion tube adapts itself to a variety of containers
- Different materials and styles suit a wide range of applications
Safely transfer liquids keeping both hands on the container you are filling. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX06040-33
- MFLX06040-35
- MFLX06040-39
- Outlet connection:
- Spout
- Spout
- Spout
- Tube ext.Ø, inlet:
- 1,8 cm (0,71")
- 2,2 cm (0,866")
- 1,6 cm (0,63")
- Tube length, inlet:
- 70 cm (27,5")
- 90 cm (35,5")
- 70 cm (27,5")
- Wetted materials:
- Polypropylene (PP)
- Polypropylene (PP)
- PVDF
- Warranty:
- 15 Months
- 15 Months
- 15 Months
Specifications
Product Information
- Katalog #Dostawca #Maximum Flow RateContainer adapterPkAvailabilityPrice
- Foot-Operated Dispensing Pump w/ Spout, PP, 26 L/min, 27,5" L inlet; 1/eaFoot-Operated Dispensing Pump w/ Spout, PP, 26 L/min, 27,5" L inlet; 1/ea06040-3326 L/min (6,9 gal./min)Universal bung for 1-3/16" to 2-3/4" (30 to 70 mm) openings1Log in for availability1 * 1 SZT850 złSupplier Article Number: 06040-33Local Article Number: MFLB06040-33Specifications:
More
Specifications
Environmentally PreferableOutlet connectionSpoutTube ext.Ø, inlet1,8 cm (0,71")Tube length, inlet70 cm (27,5")Wetted materialsPolypropylene (PP)Warranty15 Months - Foot-Operated Dispensing Pump w/ Spout, PP, 30 L/min, 35,5" L inlet; 1/eaFoot-Operated Dispensing Pump w/ Spout, PP, 30 L/min, 35,5" L inlet; 1/ea06040-3530 L/min (7,9 gal./min)Universal bung for 1-3/16" to 2-3/4" (30 to 70 mm) openings1Log in for availability1 * 1 SZT932 złSupplier Article Number: 06040-35Local Article Number: MFLB06040-35Specifications:
More
Specifications
Environmentally PreferableOutlet connectionSpoutTube ext.Ø, inlet2,2 cm (0,866")Tube length, inlet90 cm (35,5")Wetted materialsPolypropylene (PP)Warranty15 Months - Foot-Operated Dispensing Pump w/ Spout, PVDF, 20 L/min, 27,5" L inlet; 1/eaFoot-Operated Dispensing Pump w/ Spout, PVDF, 20 L/min, 27,5" L inlet; 1/ea06040-3920 L/min (5,3 gal./min)Universal bung for 1-3/16" to 2-3/4" (30 to 70 mm) openings1Log in for availability1 * 1 SZT1 270 złSupplier Article Number: 06040-39Local Article Number: MFLB06040-39Specifications:
More
Specifications
Environmentally PreferableOutlet connectionSpoutTube ext.Ø, inlet1,6 cm (0,63")Tube length, inlet70 cm (27,5")Wetted materialsPVDFWarranty15 Months