Masterflex® Union Fittings, Hose Barb, Tee, HDPE, Avantor®
About this item
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX40613-05
- MFLX30623-77
- MFLX40624-27
- MFLX30623-73
- MFLX30627-58
- MFLX40613-01
- MFLX40623-64
- MFLX40627-56
- MFLX40623-56
- MFLX30623-58
- MFLX30613-33
- MFLX30623-71
- MFLX30627-73
- MFLX30623-60
- MFLX30623-67
- MFLX30627-60
- MFLX30627-79
- MFLX90009-27
- MFLX30623-79
- MFLX30627-77
- MFLX30627-71
- MFLX30627-67
- Model:
- 40613-05
- 30623-77
- 40624-27
- 30623-73
- 30627-58
- 40613-01
- 40623-64
- 40627-56
- 40623-56
- 30623-58
- 30613-33
- 30623-71
- 30627-73
- 30623-60
- 30623-67
- 30627-60
- 30627-79
- 90009-27
- 30623-79
- 30627-77
- 30627-71
- 30627-67
- Orifice dimensions:
- 0,25"
- 0,1875"
- 0,46875"
- 0,46875"
- 0,125"
- 0,1875"
- 0,5625"
- 0,09375"
- 0,5625"
- 0,34375"
- 0,34375"
- 0,25"
- 0,125"
- 0,09375"
- Sterilisation:
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene Oxide (ETO)
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
- Ethylene oxide (ETO) only
Specifications
Product Information
- Catalog #Barb size/tubing int.Ø (port 1)Barb size/tubing int.Ø (port 2)Barb size/tubing int.Ø (port 3)Connection TypeFitting materialFlow patternAvailabilityPrice
- Fitting, HDPE, Tee, Hose Barb Unions, 1" ID; 10/PKFitting, HDPE, Tee, Hose Barb Unions, 1" ID; 10/PK1"1"1"Barbed UnionHDPET1 * 10 SZT168 złSpecifications:CE certificate
More
Specifications
Model30613-33Environmentally PreferableOrifice dimensionsSterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 5/8" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 5/8" ID; 10/PK5/8"5/8"5/8"Barbed UnionHDPET1 * 10 SZT129 złSpecifications:CE certificate
More
Specifications
Model30623-58Environmentally PreferableOrifice dimensions0,46875"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/4" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 3/4" ID; 10/PK3/4"3/4"3/4"Barbed UnionHDPET1 * 10 SZT146 złSpecifications:CE certificate
More
Specifications
Model30623-60Environmentally PreferableOrifice dimensions0,5625"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/16" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 3/16" ID; 10/PK3/16"3/16"3/16"Barbed UnionHDPET1 * 10 SZT126 złSpecifications:CE certificate
More
Specifications
Model30623-67Environmentally PreferableOrifice dimensions0,09375"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/4" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 1/4" ID; 10/PK1/4"1/4"1/4"Barbed UnionHDPET1 * 10 SZT125 złSpecifications:CE certificate
More
Specifications
Model30623-71Environmentally PreferableOrifice dimensions0,125"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 5/16" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 5/16" ID; 10/PK5/16"5/16"5/16"Barbed UnionHDPET1 * 10 SZT125 złSpecifications:CE certificate
More
Specifications
Model30623-73Environmentally PreferableOrifice dimensions0,1875"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/8" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 3/8" ID; 10/PK3/8"3/8"3/8"Barbed UnionHDPET1 * 10 SZT126 złSpecifications:CE certificate
More
Specifications
Model30623-77Environmentally PreferableOrifice dimensions0,25"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/2" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 1/2" ID; 10/PK1/2"1/2"1/2"Barbed UnionHDPET1 * 10 SZT146 złSpecifications:CE certificate
More
Specifications
Model30623-79Environmentally PreferableOrifice dimensions0,34375"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 5/8" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 5/8" ID; 100/PK5/8"5/8"5/8"Barbed UnionHDPET1 * 100 SZT663 złSpecifications:CE certificate
More
Specifications
Model30627-58Environmentally PreferableOrifice dimensions0,46875"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/4" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 3/4" ID; 100/PK3/4"3/4"3/4"Barbed UnionHDPET1 * 100 SZT725 złSpecifications:CE certificate
More
Specifications
Model30627-60Environmentally PreferableOrifice dimensions0,5625"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/16" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 3/16" ID; 100/PK3/16"3/16"3/16"Barbed UnionHDPET1 * 100 SZT586 złSpecifications:CE certificate
More
Specifications
Model30627-67Environmentally PreferableOrifice dimensions0,09375"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/4" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 1/4" ID; 100/PK1/4"1/4"1/4"Barbed UnionHDPET1 * 100 SZT620 złSpecifications:CE certificate
More
Specifications
Model30627-71Environmentally PreferableOrifice dimensions0,125"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 5/16" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 5/16" ID; 100/PK5/16"5/16"5/16"Barbed UnionHDPET1 * 100 SZT749 złSpecifications:CE certificate
More
Specifications
Model30627-73Environmentally PreferableOrifice dimensions0,1875"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/8" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 3/8" ID; 100/PK3/8"3/8"3/8"Barbed UnionHDPET1 * 100 SZT615 złSpecifications:CE certificate
More
Specifications
Model30627-77Environmentally PreferableOrifice dimensions0,25"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/2" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 1/2" ID; 100/PK1/2"1/2"1/2"Barbed UnionHDPET1 * 100 SZT696 złSpecifications:CE certificate
More
Specifications
Model30627-79Environmentally PreferableOrifice dimensions0,34375"SterilisationEthylene oxide (ETO) onlyCE certificate - Fitting, HDPE, Tee, Hose Barb Unions, 1/16" ID; 10/PKFitting, HDPE, Tee, Hose Barb Unions, 1/16" ID; 10/PK1/16"1/16"1/16"Barbed UnionHDPET1 * 10 SZT135 złSpecifications:CE certificate
More
Specifications
Model40613-01Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO)CE certificate - Fitting, HDPE, Tee, Hose Barb Unions, 1/8" ID; 10/PKFitting, HDPE, Tee, Hose Barb Unions, 1/8" ID; 10/PK1/8"1/8"1/8"Barbed UnionHDPET1 * 10 SZT103 złSpecifications:CE certificate
More
Specifications
Model40613-05Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO)CE certificate - Fitting, HDPE, Tee, Hose Barb Union, 3/32" ID; 10/PkFitting, HDPE, Tee, Hose Barb Union, 3/32" ID; 10/Pk3/32"3/32"3/32"Barbed UnionHDPET1 * 10 SZT126 złSpecifications:CE certificate
More
Specifications
Model40623-56Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO)CE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/8" ID; 10/PkFitting, HDPE, Tee, Hose Barb Union, 1/8" ID; 10/Pk1/8"1/8"1/8"Barbed UnionHDPET1 * 10 SZT108 złSpecifications:CE certificate
More
Specifications
Model40623-64Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO)CE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/16" ID; 10/PKFitting, HDPE, Tee, Hose Barb Union, 1/16" ID; 10/PK1/16"1/16"1/16"Barbed UnionHDPET1 * 10 SZT116 złSpecifications:
More
Specifications
Model40624-27Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO) - Fitting, HDPE, Tee, Hose Barb Union, 3/32" ID; 100/PkFitting, HDPE, Tee, Hose Barb Union, 3/32" ID; 100/Pk3/32"3/32"3/32"Barbed UnionHDPET1 * 100 SZT596 złSpecifications:CE certificate
More
Specifications
Model40627-56Environmentally PreferableOrifice dimensionsSterilisationEthylene Oxide (ETO)CE certificate - Fitting, HDPE, Tee, Hose Barb Union, 1/8" ID; 100/PKFitting, HDPE, Tee, Hose Barb Union, 1/8" ID; 100/PK1/8"1/8"1/8"Barbed UnionHDPET1 * 100 SZT557 złSpecifications:
More
Specifications
Model90009-27Environmentally PreferableOrifice dimensionsSterilisation