About this item
High Performance Polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.
- Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
- Ideal for general and bar code labelling on flat, smooth surfaces
- Service temperature of −40 to 160 °C (−40 to 320 °F)
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Product Information
- Catalog #Supplier #TypeModelMaterialAdhesivePrinting methodColourAvailabilityPrice
- BRDY174355174355B-488M6-80-488PolyesterPermanent acrylicWhiteLog in for availability1 * 1 Roll272 złSpecifications:
Specifications
Environmentally Preferable - BRDY174377174377B-488M6-97-488PolyesterPermanent acrylicWhiteLog in for availability1 * 1 Roll452 złSpecifications:
Specifications
Environmentally Preferable - BRDY174356174356B-488M6-81-488PolyesterPermanent acrylicWhiteLog in for availability1 * 1 Roll632 złSpecifications:
Specifications
Environmentally Preferable - BRDY174353174353B-488M6-78-488PolyesterPermanent acrylicWhiteLog in for availability1 * 1 Roll496 złSpecifications:
Specifications
Environmentally Preferable