Order Entry
Guard Column Holders
Guard Column Holders
Supplier:  ISERA
Certificates
Guard Column Holders
Supplier:  ISERA
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Guard column holder for direct coupling of 5 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
    Guard column holder for direct coupling of 5 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
    ISERAA13-000001
    AA13-000001
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 369 zł
    369 zł
    1 * 1 SZT
       369 zł
    Supplier Article Number: AA13-000001
    Local Article Number: ISERAA13-000001
    Specifications:
    Specifications
    Environmentally Preferable
  • Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
    Guard column holder for direct coupling of 10 mm cartridges to ISERA analytical columns (2 - 4,6 mm Int.Ø)
    ISERAA13-000002
    AA13-000002
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 369 zł
    369 zł
    1 * 1 SZT
       369 zł
    Supplier Article Number: AA13-000002
    Local Article Number: ISERAA13-000002
    Specifications:
    Specifications
    Environmentally Preferable
  • Guard column holder 'stand-alone' for 5 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    Guard column holder 'stand-alone' for 5 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    ISERAA13-000003
    AA13-000003
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 369 zł
    369 zł
    1 * 1 SZT
       369 zł
    Supplier Article Number: AA13-000003
    Local Article Number: ISERAA13-000003
    Specifications:
    Specifications
    Environmentally Preferable
  • Guard column holder 'stand-alone' for 10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    Guard column holder 'stand-alone' for 10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    ISERAA13-000004
    AA13-000004
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 369 zł
    369 zł
    1 * 1 SZT
       369 zł
    Supplier Article Number: AA13-000004
    Local Article Number: ISERAA13-000004
    Specifications:
    Specifications
    Environmentally Preferable
  • Guard column holder (semi-preparative, 'stand-alone') for 7,8×10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    Guard column holder (semi-preparative, 'stand-alone') for 7,8×10 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    ISERAA13-000005
    AA13-000005
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 340 zł
    1 340 zł
    1 * 1 SZT
       1 340 zł
    Supplier Article Number: AA13-000005
    Local Article Number: ISERAA13-000005
    Specifications:
    Specifications
    Environmentally Preferable
  • Guard column holder (preparative, 'stand-alone') for 10×21,2 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    Guard column holder (preparative, 'stand-alone') for 10×21,2 mm cartridges, ¹/₁₆" (10-32) thread on both sides
    ISERAA13-000006
    AA13-000006
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 1 710 zł
    1 710 zł
    1 * 1 SZT
       1 710 zł
    Supplier Article Number: AA13-000006
    Local Article Number: ISERAA13-000006
    Specifications:
    Specifications
    Environmentally Preferable
  • UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
    UHPLC/HPLC guard column holder for direct coupling of 5 mm guard cartridges (zero dead volume) to analytical columns, suitable for XELA and XCORE cartridges, can be mounted on analytical columns of most manufacturers
    ISERAA13-000007
    AA13-000007
    1
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 770 zł
    770 zł
    1 * 1 SZT
       770 zł
    Supplier Article Number: AA13-000007
    Local Article Number: ISERAA13-000007
    Specifications:
    Specifications
    Environmentally Preferable