Drukuj
Liebherr, Kombinowane chłodziarko-zamrażarki laboratoryjne z Comfort Electronic, Mediline
Dostawca: Liebherr
471-0696EA 11930 PLN
471-0696
Liebherr, Kombinowane chłodziarko-zamrażarki laboratoryjne z Comfort Electronic, Mediline
Chłodziarki/Lodówki
Combined laboratory refrigerator and freezer, with independent dual 'Comfort' refrigeration control and compression systems for both compartments so that the refrigerator and freezer sections can be operated separately.
  • Electronic controllers (one for fridge, one for freezer)
  • Self closing doors with ergonomic handles and door locks
  • RS485 interface
  • Three alarm event memory, door and temperature alarms can be customised to suit end user requirements
  • Dual compressor and dual controller
Temperature range: 3 to 8 °C (fridge); –9 to –30 °C (freezer)
Informacje do zamówienia: An optional product temperature probe is available and allows the controller to display the air and the product temperatures. Please contact your local Avantor office for details. Also please note that a spark-free version of this unit is available, LCexv 4010, please enquire for details.